- Use of stains to detect fingermarks.
Use of stains to detect fingermarks.
Biotechnic & histochemistry : official publication of the Biological Stain Commission (2010-02-04)
A Becue, S Moret, C Champod, P Margot
PMID20121464
ABSTRACT
Detection of fingermarks at a crime scene or on related items is of prime interest for forensic investigators, mainly for identification purposes. Most of the fingermarks are invisible to the naked eye, however. The application of detection techniques is required to establish visual contrast between the secretion residue and the underlying substrate. We give here a review of the field related to the concept of using stains to detect fingermarks. A distinction has been made between the physically driven classical detection techniques, the chemically driven ones, and those based on nanostructured materials, an emerging field in forensic science.