Merck
  • Detecting cells on the surface of a silver electrode quartz crystal microbalance using plasma treatment and graft polymerization.
Detecting cells on the surface of a silver electrode quartz crystal microbalance using plasma treatment and graft polymerization.
Colloids and surfaces. B, Biointerfaces (2009-06-24)
Hung-Che Chou, Tsong-Rong Yan, Ko-Shao Chen
ABSTRACT
This paper utilizes a silver electrode quartz crystal microbalance (QCM) mass sensor to detect the physiology of cells. This study also investigates the plasma surface modification of silver electrode QCMs through deposition of hexamethyldisilazane (HMDSZ) films as a protection film. To improve the cell growth, this paper also performs post-treatments by surface-grafting acrylic acid (AAc), acrylamide (AAm), and oxygen plasma treatment onto the QCM electrodes. Experimental results indicate that plasma deposition is a useful technique to protect the surface of silver electrodes. This technique extends the unpeeling time of silver electrodes from 1 to 7 days. The hydrophilic silver electrode QCM surface modified by AAm exhibited a better storage time effect than other post-treatments.
Sigma-Aldrich
Tris[N,N-bis(trimethylsilyl)amide]yttrium
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide, 97%
Sigma-Aldrich
Tris[N,N-bis(trimethylsilyl)amide]samarium(III), 98%
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide, 95%
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 0.5 M in toluene
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in THF
Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Hexamethyldisilazane, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 40% in THF
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1 M in toluene
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1 M in tert-butyl methyl ether
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide, 95%
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 0.6 M in toluene
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 1.0 M in THF
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in hexanes
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1.0 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 0.5 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.5 M in THF
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1 M in THF