Merck
  • Reduction of 1,4-dien-3-one steroids with LiAl2H4 or NaB2H4: stereospecific deuterium-labeling at the c-1alpha position of a 4-en-3-one steroid.
Reduction of 1,4-dien-3-one steroids with LiAl2H4 or NaB2H4: stereospecific deuterium-labeling at the c-1alpha position of a 4-en-3-one steroid.
Chemical & pharmaceutical bulletin (2006-04-06)
Mitsuteru Numazawa, Wakako Handa
ABSTRACT
Reduction of a double bond at C-1 of 1,4-dien-3-one steroids 7 and 8 with LiAl2H4 in THF or NaB2H4 in MeOH and H2O gave stereospecifically [1alpha-2H]-labeled 4-en-3-one steroids 9 and 10, respectively. When the deuterated solvents, MeO2H and 2H2O, were used for the reaction of steroid 8 with NaB2H4, [1alpha,2xi-2H2]-labeled compound 10 was produced. This indicates that the reaction proceeds through the initial hydride attack at the C-1alpha position, followed by ketonization of the 2-en-3-ol intermediate.
Sigma-Aldrich
Lithium aluminum hydride solution, 2.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride, powder, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric), tablet (5 g each)
Sigma-Aldrich
Lithium aluminum hydride, pellets, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric)
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride, hydrogen-storage grade
Sigma-Aldrich
Lithium aluminum hydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Lithium aluminum hydride solution, 2.3 M in 2-methyltetrahydrofuran