Merck
  • alpha-Fetoprotein as a modulator of the pro-inflammatory response of human keratinocytes.
alpha-Fetoprotein as a modulator of the pro-inflammatory response of human keratinocytes.
British journal of pharmacology (2009-09-30)
A I Potapovich, S Pastore, V A Kostyuk, D Lulli, V Mariani, C De Luca, E I Dudich, L G Korkina
ABSTRACT
The immunomodulatory effects of alpha-fetoprotein (AFP) on lymphocytes and macrophages have been described in vitro and in vivo. Recombinant forms of human AFP have been proposed as potential therapeutic entities for the treatment of autoimmune diseases. We examined the effects of embryonic and recombinant human AFP on the spontaneous, UVA- and cytokine-induced pro-inflammatory responses of human keratinocytes. Cultures of primary and immortalized human keratinocytes (HaCaT) and human blood T lymphocytes were used. The effects of AFP on cytokine expression were studied by bioplexed elisa and quantitative reverse transcriptase polymerase chain reaction assay. Kinase and nuclear factor kappa B (NFkappaB) phosphorylation were quantified by intracellular elisa. Nuclear activator protein 1 and NFkappaB DNA binding activity was measured by specific assays. Nitric oxide and H(2)O(2) production and redox status were assessed by fluorescent probe and biochemical methods. All forms of AFP enhanced baseline expression of cytokines, chemokines and growth factors. AFP dose-dependently increased tumour necrosis factor alpha-stimulated granulocyte macrophage colony stimulating factor and interleukin 8 expression and decreased tumour necrosis factor alpha-induced monocyte chemotactic protein 1 and IP-10 (interferon gamma-produced protein of 10 kDa) expression. AFP induced a marked activator protein 1 activation in human keratinocytes. AFP also increased H(2)O(2) and modulated nitrite/nitrate levels in non-stimulated keratinocytes whereas it did not affect these parameters or cytokine release from UVA-stimulated cells. Phosphorylation of extracellular signal-regulated kinase (ERK1/2) and Akt1 but not NFkappaB was activated by AFP alone or by its combination with UVA. Exogenous AFP induces activation of human keratinocytes, with de novo expression of a number of pro-inflammatory mediators and modulation of their pro-inflammatory response to cytokines or UVA. AFP may modulate inflammatory events in human skin.
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 70 purifications
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 10 purifications
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077 g/mL
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 350 purifications
Sigma-Aldrich
Plasma, from mouse
Sigma-Aldrich
Plasma, from rat
Sigma-Aldrich
Plasma, from chicken
Sigma-Aldrich
Plasma, from bovine
Sigma-Aldrich
Plasma, from pig
Sigma-Aldrich
Plasma, from human