Merck
  • Cloning, expression, and characterization of a peculiar choline-binding beta-galactosidase from Streptococcus mitis.
Cloning, expression, and characterization of a peculiar choline-binding beta-galactosidase from Streptococcus mitis.
Applied and environmental microbiology (2009-07-28)
Susana Campuzano, Beatriz Serra, Daniel Llull, José L García, Pedro García
ABSTRACT
A Streptococcus mitis genomic DNA fragment carrying the SMT1224 gene encoding a putative beta-galactosidase was identified, cloned, and expressed in Escherichia coli. This gene encodes a protein 2,411 amino acids long with a predicted molecular mass of 268 kDa. The deduced protein contains an N-terminal signal peptide and a C-terminal choline-binding domain consisting of five consensus repeats, which facilitates the anchoring of the secreted enzyme to the cell wall. The choline-binding capacity of the protein facilitates its purification using DEAE-cellulose affinity chromatography, although its complete purification was achieved by constructing a His-tagged fusion protein. The recombinant protein was characterized as a monomeric beta-galactosidase showing a specific activity of around 2,500 U/mg of protein, with optimum temperature and pH ranges of 30 to 40 degrees C and 6.0 to 6.5, respectively. Enzyme activity is not inhibited by glucose, even at 200 mM, and remains highly stable in solution or immobilized at room temperature in the absence of protein stabilizers. In S. mitis, the enzyme was located attached to the cell surface, but a significant activity was also detected in the culture medium. This novel enzyme represents the first beta-galactosidase having a modular structure with a choline-binding domain, a peculiar property that can also be useful for some biotechnological applications.
Product Description
Supelco
Whatman® cellulose chromatography papers, 1 Chr roll, 1.5 inch x 300 feet, 1/pk
Whatman® cellulose chromatography papers, 3 mm blotting paper, diam. 25 mm
Whatman® cellulose chromatography papers, 3 mm blotting paper, diam. 24 mm
Whatman® cellulose chromatography papers, 4 CHR Roll, W × L 20 mm × 100 m
Whatman® cellulose chromatography papers, 17-chr, W × L 25 mm × 220 mm
Whatman® cellulose chromatography papers, 3MM Chr roll, 10 cm x 100 m, 1/pk
Whatman® cellulose chromatography papers, 2727 circles, diam. 185 mm
Whatman® cellulose chromatography papers, 2727-CHR Sheets, W × L 190 mm × 190 mm
Whatman® cellulose chromatography papers, 31ET CHR Chroma Reel, W × L 15 cm × 100 m
Whatman® cellulose chromatography papers, 3MM CHR, W × L 23 cm × 100 m, pkg of 1 roll
Whatman® cellulose chromatography papers, 3MM CHR, W × L 11 cm × 14 cm, pkg of 100 ea
Whatman® cellulose chromatography papers, 3MM CHR, W × L 18 cm × 34 cm, pkg of 100 ea
Whatman® cellulose chromatography papers, 1 CHR Rol, W × L 20 mm × 100 m, pkg of 1 roll
Whatman® cellulose chromatography papers, 3MM CHR, W × L 4 in. × 5.25 in., pkg of 100 ea
Whatman® cellulose chromatography papers, 1-chr, W × L 580 mm × 680 mm
Whatman® cellulose chromatography papers, 1 CHR Rol, W × L 40 mm × 100 m, pkg of 1 roll
Whatman® cellulose chromatography papers, 4 Chr sheets, 46 x 57 cm, 100/pk
Whatman® cellulose chromatography papers, 3MM CHR, W × L 12 cm × 14 cm, pkg of 100 ea
Whatman® cellulose chromatography papers, SG81 Sheets, W × L 460 mm × 570 mm
Whatman® cellulose chromatography papers, 3MM CHR, W × L 27 cm × 100 m, pkg of 1 roll
Whatman® cellulose chromatography papers, 31ET sheets, W × L 20 mm × 50 mm
Whatman® cellulose chromatography papers, 17-chr, W × L 2.5 cm × 30 m
Whatman® cellulose chromatography papers, 1 CHR Rol, W × L 50 mm × 100 m, pkg of 1 roll
Whatman® cellulose chromatography papers, 1 CHR Sheets, W × L 100 mm × 300 mm, pkg of 100 ea
Whatman® cellulose chromatography papers, 3MM CHR, W × L 26 cm × 41 cm, pkg of 100 ea
Whatman® cellulose chromatography papers, 2727 sheets, W × L 580 mm × 600 mm
Whatman® cellulose chromatography papers, 1 CHR Sheets, W × L 250 mm × 250 mm, pkg of 100 ea
Whatman® cellulose chromatography papers, 2 Chr sheets, 46 x 57 cm, 100/pk
Whatman® cellulose chromatography papers, 1 Chr sheets, 46 x 57 cm, 100/pk
Whatman® cellulose chromatography papers, 3MM CHR, W × L 31.5 cm × 35.5 cm, pkg of 100 ea