Merck
Thioredoxin is a metabolic rheostat controlling regulatory B cells.
Nature immunology (2024-03-30)
Hannah F Bradford, Thomas C R McDonnell, Alexander Stewart, Andrew Skelton, Joseph Ng, Zara Baig, Franca Fraternali, Deborah Dunn-Walters, David A Isenberg, Adnan R Khan, Claudio Mauro, Claudia Mauri
ABSTRACT
Metabolic programming is important for B cell fate, but the bioenergetic requirement for regulatory B (Breg) cell differentiation and function is unknown. Here we show that Breg cell differentiation, unlike non-Breg cells, relies on mitochondrial electron transport and homeostatic levels of reactive oxygen species (ROS). Single-cell RNA sequencing analysis revealed that TXN, encoding the metabolic redox protein thioredoxin (Trx), is highly expressed by Breg cells, unlike Trx inhibitor TXNIP which was downregulated. Pharmacological inhibition or gene silencing of TXN resulted in mitochondrial membrane depolarization and increased ROS levels, selectively suppressing Breg cell differentiation and function while favoring pro-inflammatory B cell differentiation. Patients with systemic lupus erythematosus (SLE), characterized by Breg cell deficiencies, present with B cell mitochondrial membrane depolarization, elevated ROS and fewer Trx+ B cells. Exogenous Trx stimulation restored Breg cells and mitochondrial membrane polarization in SLE B cells to healthy B cell levels, indicating Trx insufficiency underlies Breg cell impairment in patients with SLE.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Buthionine-sulfoximine, ≥97% (TLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Goat Anti-Human IgM+IgG+IgA Antibody, F(ab′)2, 0.5 mg/mL, Chemicon®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MitoTEMPO, ≥98% (HPLC)