Merck
  • Human pallial MGE-type GABAergic interneuron cell therapy for chronic focal epilepsy.
Human pallial MGE-type GABAergic interneuron cell therapy for chronic focal epilepsy.
Cell stem cell (2023-10-07)
Marina Bershteyn, Sonja Bröer, Mansi Parekh, Yves Maury, Steven Havlicek, Sonja Kriks, Luis Fuentealba, Seonok Lee, Robin Zhou, Geetha Subramanyam, Meliz Sezan, Eric Steven Sevilla, Whitney Blankenberger, Julien Spatazza, Li Zhou, Hubert Nethercott, David Traver, Philip Hampel, Hannah Kim, Michael Watson, Naomi Salter, Anastasia Nesterova, Wai Au, Arnold Kriegstein, Arturo Alvarez-Buylla, John Rubenstein, Gautam Banik, Alessandro Bulfone, Catherine Priest, Cory R Nicholas
ABSTRACT
Mesial temporal lobe epilepsy (MTLE) is the most common focal epilepsy. One-third of patients have drug-refractory seizures and are left with suboptimal therapeutic options such as brain tissue-destructive surgery. Here, we report the development and characterization of a cell therapy alternative for drug-resistant MTLE, which is derived from a human embryonic stem cell line and comprises cryopreserved, post-mitotic, medial ganglionic eminence (MGE) pallial-type GABAergic interneurons. Single-dose intrahippocampal delivery of the interneurons in a mouse model of chronic MTLE resulted in consistent mesiotemporal seizure suppression, with most animals becoming seizure-free and surviving longer. The grafted interneurons dispersed locally, functionally integrated, persisted long term, and significantly reduced dentate granule cell dispersion, a pathological hallmark of MTLE. These disease-modifying effects were dose-dependent, with a broad therapeutic range. No adverse effects were observed. These findings support an ongoing phase 1/2 clinical trial (NCT05135091) for drug-resistant MTLE.
Sigma-Aldrich
Anti-Doublecortin Antibody, serum, from guinea pig
Sigma-Aldrich
Anti-NeuN Antibody, from chicken, purified by affinity chromatography
Sigma-Aldrich
Anti-Nuclei Antibody, clone 235-1, clone 235-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-MAFB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Neuropeptide Y Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Oligodendrocyte transcription factor 2 (OLIG2 Antibody), serum, from guinea pig