Merck
Rescue of glutaric aciduria type I in mice by liver-directed therapies.
Science translational medicine (2023-04-19)
Mercedes Barzi, Collin G Johnson, Tong Chen, Ramona M Rodriguiz, Madeline Hemmingsen, Trevor J Gonzalez, Alan Rosales, James Beasley, Cheryl K Peck, Yunhan Ma, Ashlee R Stiles, Timothy C Wood, Raquel Maeso-Diaz, Anna Mae Diehl, Sarah P Young, Jeffrey I Everitt, William C Wetsel, William R Lagor, Beatrice Bissig-Choisat, Aravind Asokan, Areeg El-Gharbawy, Karl-Dimiter Bissig
ABSTRACT
Glutaric aciduria type I (GA-1) is an inborn error of metabolism with a severe neurological phenotype caused by the deficiency of glutaryl-coenzyme A dehydrogenase (GCDH), the last enzyme of lysine catabolism. Current literature suggests that toxic catabolites in the brain are produced locally and do not cross the blood-brain barrier. In a series of experiments using knockout mice of the lysine catabolic pathway and liver cell transplantation, we uncovered that toxic GA-1 catabolites in the brain originated from the liver. Moreover, the characteristic brain and lethal phenotype of the GA-1 mouse model was rescued by two different liver-directed gene therapy approaches: Using an adeno-associated virus, we replaced the defective Gcdh gene or we prevented flux through the lysine degradation pathway by CRISPR deletion of the aminoadipate-semialdehyde synthase (Aass) gene. Our findings question the current pathophysiological understanding of GA-1 and reveal a targeted therapy for this devastating disorder.
Sigma-Aldrich
Anti-GCDH antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Anti-FAH, IgG fraction of antiserum
Sigma-Aldrich
Anti-AASS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2