Merck
  • High genetic stability of dengue virus propagated in MRC-5 cells as compared to the virus propagated in vero cells.
High genetic stability of dengue virus propagated in MRC-5 cells as compared to the virus propagated in vero cells.
PloS one (2008-03-20)
Chia-Chyi Liu, Shiang-Chi Lee, Michael Butler, Suh-Chin Wu
ABSTRACT
This work investigated the replication kinetics of the four dengue virus serotypes (DEN-1 to DEN-4), including dengue virus type 4 (DEN-4) recovered from an infectious cDNA clone, in Vero cells and in MRC-5 cells grown on Cytodex 1 microcarriers. DEN-1 strain Hawaii, DEN-2 strain NGC, DEN-3 strain H-87, and DEN-4 strain H-241 , and DEN-4 strain 814669 derived from cloned DNA, were used to infect Vero cells and MRC-5 cells grown in serum-free or serum-containing microcarrier cultures. Serum-free and serum-containing cultures were found to yield comparable titers of these viruses. The cloned DNA-derived DEN-4 started genetically more homogeneous was used to investigate the genetic stability of the virus propagated in Vero cells and MRC-5 cells. Sequence analysis revealed that the DEN-4 propagated in MRC-5 cells maintained a high genetic stability, compared to the virus propagated in Vero cells. Amino acid substitutions of Gly(104)Cys and Phe(108)Ile were detected at 70%, 60%, respectively, in the envelope (E) protein of DEN-4 propagated in Vero cells, whereas a single mutation of Glu(345)Lys was detected at 50% in E of the virus propagated in MRC-5 cells. Sequencing of multiple clones of three separate DNA fragments spanning 40% of the genome also indicated that DEN-4 propagated in Vero cells contained a higher number of mutations than the virus growing in MRC-5 cells. Although Vero cells yielded a peak virus titer approximately 1 to 17 folds higher than MRC-5 cells, cloned DEN-4 from MRC-5 cells maintained a greater stability than the virus from Vero cells. Serum-free microcarrier cultures of MRC-5 cells offer a potentially valuable system for the large-scale production of live-attenuated DEN vaccines.
Product Description
SAFC
Minimum Essential Medium, with Earle′s salts, with 2.0 mM L-glutamine, with 20.0 mM HEPES, with non-essential amino acids, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 2.0 mM L-glutamine, without sodium bicarbonate, dry powder, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 2.0 mM L-glutamine, with non-essential amino acids, without sodium bicarbonate, dry powder, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 2.0 mM L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 25mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with non-essential amino acids, without L-glutamine, liquid, sterile-filtered, suitable for cell culture