Merck
  • Protein and RNA ADP-ribosylation detection is influenced by sample preparation and reagents used.
Protein and RNA ADP-ribosylation detection is influenced by sample preparation and reagents used.
Life science alliance (2022-11-12)
Lisa Weixler, Nonso Josephat Ikenga, Jim Voorneveld, Gülcan Aydin, Timo Mhr Bolte, Jeffrey Momoh, Mareike Bütepage, Alexandra Golzmann, Bernhard Lüscher, Dmitri V Filippov, Roko Žaja, Karla Lh Feijs
ABSTRACT
The modification of substrates with ADP-ribose (ADPr) is important in, for example, antiviral immunity and cancer. Recently, several reagents were developed to detect ADP-ribosylation; however, it is unknown whether they recognise ADPr, specific amino acid-ADPr linkages, or ADPr with the surrounding protein backbone. We first optimised methods to prepare extracts containing ADPr-proteins and observe that depending on the amino acid modified, the modification is heatlabile. We tested the reactivity of available reagents with diverse ADP-ribosylated protein and RNA substrates and observed that not all reagents are equally suited for all substrates. Next, we determined cross-reactivity with adenylylated RNA, AMPylated proteins, and metabolites, including NADH, which are detected by some reagents. Lastly, we analysed ADP-ribosylation using confocal microscopy, where depending on the fixation method, either mitochondrion, nucleus, or nucleolus is stained. This study allows future work dissecting the function of ADP-ribosylation in cells, both on protein and on RNA substrates, as we optimised sample preparation methods and have defined the reagents suitable for specific methods and substrates.
Sigma-Aldrich
Anti-mono- ADP-ribose binding reagent, from Escherichia coli