Merck
  • Beyond a PPR-RNA recognition code: Many aspects matter for the multi-targeting properties of RNA editing factor PPR56.
Beyond a PPR-RNA recognition code: Many aspects matter for the multi-targeting properties of RNA editing factor PPR56.
PLoS genetics (2023-08-21)
Yingying Yang, Kira Ritzenhofen, Jessica Otrzonsek, Jingchan Xie, Mareike Schallenberg-Rüdinger, Volker Knoop
ABSTRACT
The mitochondrial C-to-U RNA editing factor PPR56 of the moss Physcomitrium patens is an RNA-binding pentatricopeptide repeat protein equipped with a terminal DYW-type cytidine deaminase domain. Transferred into Escherichia coli, PPR56 works faithfully on its two native RNA editing targets, nad3eU230SL and nad4eU272SL, and also converts cytidines into uridines at over 100 off-targets in the bacterial transcriptome. Accordingly, PPR56 is attractive for detailed mechanistic studies in the heterologous bacterial setup, allowing for scoring differential RNA editing activities of many target and protein variants in reasonable time. Here, we report (i) on the effects of numerous individual and combined PPR56 protein and target modifications, (ii) on the spectrum of off-target C-to-U editing in the bacterial background transcriptome for PPR56 and two variants engineered for target re-direction and (iii) on combinations of targets in tandem or separately at the 5'- and 3'-ends of large mRNAs. The latter experimentation finds enhancement of RNA editing at weak targets in many cases, including cox3eU290SF as a new candidate mitogenome target. We conclude that C-to-U RNA editing can be much enhanced by transcript features also outside the region ultimately targeted by PPRs of a plant editing factor, possibly facilitated by its enrichment or scanning along transcripts.
Sigma-Aldrich
Rosetta Competent Cells - Novagen, Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
10MG
Usually ships in 2 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
$30.70
To order products, please contact your local dealer.