Merck
  • Determination of the polyphenolic content of berry juices using focusing-modulated comprehensive two-dimensional liquid chromatography coupled to mass spectrometry detection.
Determination of the polyphenolic content of berry juices using focusing-modulated comprehensive two-dimensional liquid chromatography coupled to mass spectrometry detection.
Analytical and bioanalytical chemistry (2022-07-15)
Katia Arena, Francesco Cacciola, Natalizia Miceli, Maria Fernanda Taviano, Emilia Cavò, Robert E Murphy, Paola Dugo, Luigi Mondello
ABSTRACT
In this work, a comprehensive two-dimensional liquid chromatography system, comprised of a ZIC-HILIC and C18 columns in the first and second dimensions, respectively, was tuned and employed for attaining high resolution profiles of the polyphenolic pattern in seven commercial berry juices. The developed HILIC × RP-LC method was validated in terms of linearity range, correlation coefficients, limit of detection, limit of quantification, precision (intra- and inter-day), and recovery. A total of 104 polyphenolic compounds belonging to different chemical classes (hydroxybenzoic and cinnamic acid derivatives, flavone glycosides, flavonols, flavonol glycosides, dihydroflavonols, and anthocyanin glycosides) have been characterized and quantified in the juices investigated. Despite the constituents being similar, a notable quantitative variation among the analyzed berry species was observed. Elderberry contained the highest amount of polyphenols (918 ± 1.10 mg 100 mL-1), followed by chokeberry (516 ± 0.08 mg 100 mL-1). On the other hand, raspberry contained the lowest amount (104 ± 1.21 mg 100 mL-1). Further, total phenolic, flavonoid, and anthocyanin contents were determined spectrophotometrically, yielding consistent results. The free-radical scavenging activity (DPPH test) and reducing power of the juices, expressed as IC50 (μL mL-1) and mg ASE mL-1, varied from 2.79 ± 0.03 (honeyberry) to 31.66 ± 0.02 (blueberry) and from 1.71 ± 0.01 (blueberry) to 8.89 ± 0.12 (chokeberry), respectively. Such a ZIC-HILIC × C18 platform based on focusing modulation, never employed so far for berry juices, showed a remarkable separation capability with high values of corrected peak capacity (up to 1372) and orthogonality (Ao up to 0.80), thus providing a great applicability to be advantageously employed for other complex food samples.
Product Description
Cyanidin 3-glucoside chloride, phyproof® Reference Substance
Supelco
Chlorogenic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 4.6 mm, pkg of 3 ea
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Taxifolin, analytical standard
Supelco
Quercetin 3-glucoside, analytical standard
Supelco
Luteolin 7-glucoside, analytical standard
Supelco
SeQuant® ZIC-HILIC (3.5µm,100Å) 150 x 1 mm, suitable for HPLC
USP
Luteolin 7-O-Glucoside, United States Pharmacopeia (USP) Reference Standard