Merck
  • Expanding the prostate cancer cell line repertoire with ACRJ-PC28, an AR-negative neuroendocrine cell line derived from an African-Caribbean patient.
Expanding the prostate cancer cell line repertoire with ACRJ-PC28, an AR-negative neuroendocrine cell line derived from an African-Caribbean patient.
Cancer research communications (2023-01-17)
Henkel Valentine, William Aiken, Belinda Morrison, Ziran Zhao, Holly Fowle, Jason S Wasserman, Elon Thompson, Warren Chin, Mark Young, Shannique Clarke, Denise Gibbs, Sharon Harrison, Wayne McLaughlin, Tim Kwok, Fang Jin, Kerry S Campbell, Anelia Horvath, Rory Thompson, Norman H Lee, Yan Zhou, Xavier Graña, Camille Ragin, Simone Badal
ABSTRACT
Prostate cell lines from diverse backgrounds are important to addressing disparities in prostate cancer (PCa) incidence and mortality rates among Black men. ACRJ-PC28 was developed from a transrectal needle biopsy and established via inactivation of the CDKN2A locus and simultaneous expression of human telomerase. Characterization assays included growth curve analysis, immunoblots, IHC, 3D cultures, immunofluorescence imaging, confocal microscopy, flow cytometry, WGS, and RNA-Seq. ACRJ-PC28 has been passaged more than 40 times in vitro over 10 months with a doubling time of 45 hours. STR profiling confirmed the novelty and human origin of the cell line. RNA-Seq confirmed the expression of prostate specific genes alpha-methylacyl-CoA racemase (AMACR) and NKX3.1 and Neuroendocrine specific markers synaptophysin (SYP) and enolase 2 (ENO2) and IHC confirmed the presence of AMACR. Immunoblots indicated the cell line is of basal-luminal type; expresses p53 and pRB and is AR negative. WGS confirmed the absence of exonic mutations and the presence of intronic variants that appear to not affect function of AR, p53, and pRB. RNA-Seq data revealed numerous TP53 and RB1 mRNA splice variants and the lack of AR mRNA expression. This is consistent with retention of p53 function in response to DNA damage and pRB function in response to contact inhibition. Soft agar anchorage-independent analysis indicated that the cells are transformed, confirmed by principal component analysis (PCA) where ACRJ-PC28 cells cluster alongside other PCa tumor tissues, yet was distinct. The novel methodology described should advance prostate cell line development, addressing the disparity in PCa among Black men.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
apo-Transferrin human, powder, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(±)-Epinephrine hydrochloride
Sigma-Aldrich
TGF- α Protein, Human Recombinant Animal Free, >98% (SDS-PAGE and HPLC), <0.1 μg/mg protein endotoxin (<1EU/μg)