Merck
Development of immortalized hepatocyte-like cells from hMSCs.
Methods in molecular biology (Clifton, N.J.) (2011-12-15)
Adisak Wongkajornsilp, Khanit Sa-Ngiamsuntorn, Suradej Hongeng
ABSTRACT
Clones of hepatocyte-like cells were reproducibly generated from human mesenchymal stem cells immortalized with a combined transduction of both Bmi-1 and TERT genes. These hepatocyte-like cells contained selective markers and several functional properties of hepatocytes, yet still carried proliferative potential. These cells had cuboidal morphology and arranged themselves as cord-like structure in culture. The cloned cells deposited glycogen and actively synthesized albumin. The basal expressions of CYP450 isozymes was observed, albeit only 10-20% that of primary hepatocytes. These expressions were promptly increased upon the addition of rifampicin, a known enzyme inducer. These hepatocyte-like cells may serve as a close alternative to the use of primary hepatocytes for in vitro studies.
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Hepatocyte Growth Factor Protein, Recombinant human, The Human Hepatocyte Growth Factor (or HGF protein) is also known as Scatter Factor & Hepatopoeitin A.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Phenobarbital sodium salt
Sigma-Aldrich
Epidermal Growth Factor Protein, Human recombinant, The Epidermal Growth Factor (EGF) is a polypeptide growth factor which stimulates the proliferation of a wide range of epidermal & epithelial cells.
Sigma-Aldrich
Omeprazole, solid