Merck
  • Human PSCs determine the competency of cerebral organoid differentiation via FGF signaling and epigenetic mechanisms.
Human PSCs determine the competency of cerebral organoid differentiation via FGF signaling and epigenetic mechanisms.
iScience (2022-10-04)
Hirosato Ideno, Kent Imaizumi, Hiroko Shimada, Tsukasa Sanosaka, Akisa Nemoto, Jun Kohyama, Hideyuki Okano
ABSTRACT
Various culture methods have been developed for maintaining human pluripotent stem cells (PSCs). These PSC maintenance methods exhibit biased differentiation; for example, feeder-dependent PSCs efficiently yield cerebral organoids, but it is difficult to generate organoids from feeder-free PSCs. It remains unknown how PSC maintenance conditions affect differentiation. In this study, we identified fibroblast growth factor (FGF) signaling in feeder-free PSC maintenance as a key factor that determines the differentiation toward cerebral organoids. The inhibition of FGF signaling in feeder-free PSCs rescued organoid generation to the same level in feeder-dependent cultures. FGF inhibition induced DNA methylation at the WNT5A locus, and this epigenetic change suppressed the future activation of non-canonical Wnt signaling after differentiation, leading to reliable cerebral organoid generation. This study underscores the importance of PSC culture conditions for directed differentiation into cerebral organoids, and the epigenetic status regulated by FGF signaling is involved in the underlying mechanisms.
Sigma-Aldrich
Tankyrase1/2 Inhibitor, XAV939, The Tankyrase1/2 Inhibitor, XAV939 controls the biological activity of Tankyrase1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Anti-N-Cadherin antibody, Mouse monoclonal, clone GC-4, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-β-Tubulin III antibody produced in mouse, clone SDL.3D10, ascites fluid
Sigma-Aldrich
IWP-2, ≥98% (HPLC)
Sigma-Aldrich
Anti-Brn-3a Antibody, POU-domain protein, clone 5A3.2, culture supernatant, clone 5A3.2, Chemicon®