Merck
  • Melanoma-Secreted Amyloid Beta Suppresses Neuroinflammation and Promotes Brain Metastasis.
Melanoma-Secreted Amyloid Beta Suppresses Neuroinflammation and Promotes Brain Metastasis.
Cancer discovery (2022-03-10)
Kevin Kleffman, Grace Levinson, Indigo V L Rose, Lili M Blumenberg, Sorin A A Shadaloey, Avantika Dhabaria, Eitan Wong, Francisco Galán-Echevarría, Alcida Karz, Diana Argibay, Richard Von Itter, Alfredo Floristán, Gillian Baptiste, Nicole M Eskow, James A Tranos, Jenny Chen, Eleazar C Vega Y Saenz de Miera, Melissa Call, Robert Rogers, George Jour, Youssef Zaim Wadghiri, Iman Osman, Yue-Ming Li, Paul Mathews, Ronald B DeMattos, Beatrix Ueberheide, Kelly V Ruggles, Shane A Liddelow, Robert J Schneider, Eva Hernando
ABSTRACT
Brain metastasis is a significant cause of morbidity and mortality in multiple cancer types and represents an unmet clinical need. The mechanisms that mediate metastatic cancer growth in the brain parenchyma are largely unknown. Melanoma, which has the highest rate of brain metastasis among common cancer types, is an ideal model to study how cancer cells adapt to the brain parenchyma. Our unbiased proteomics analysis of melanoma short-term cultures revealed that proteins implicated in neurodegenerative pathologies are differentially expressed in melanoma cells explanted from brain metastases compared with those derived from extracranial metastases. We showed that melanoma cells require amyloid beta (Aβ) for growth and survival in the brain parenchyma. Melanoma-secreted Aβ activates surrounding astrocytes to a prometastatic, anti-inflammatory phenotype and prevents phagocytosis of melanoma by microglia. Finally, we demonstrate that pharmacologic inhibition of Aβ decreases brain metastatic burden. Our results reveal a novel mechanistic connection between brain metastasis and Alzheimer's disease, two previously unrelated pathologies; establish Aβ as a promising therapeutic target for brain metastasis; and demonstrate suppression of neuroinflammation as a critical feature of metastatic adaptation to the brain parenchyma. This article is highlighted in the In This Issue feature, p. 1171.
Sigma-Aldrich
Anti-Amyloid β40 Antibody, clone G2-10, clone G2-10, from mouse
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Iba1/AIF1 Antibody, clone 20A12.1, from mouse
Sigma-Aldrich
Anti-XPNPEP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution