Merck
  • Rox, a Rifamycin Resistance Enzyme with an Unprecedented Mechanism of Action.
Rox, a Rifamycin Resistance Enzyme with an Unprecedented Mechanism of Action.
Cell chemical biology (2018-02-06)
Kalinka Koteva, Georgina Cox, Jayne K Kelso, Matthew D Surette, Haley L Zubyk, Linda Ejim, Peter Stogios, Alexei Savchenko, Dan Sørensen, Gerard D Wright
ABSTRACT
Rifamycin monooxygenases (Rox) are present in a variety of environmental bacteria and are associated with decomposition of the clinically utilized antibiotic rifampin. Here we report the structure and function of a drug-inducible rox gene from Streptomyces venezuelae, which encodes a class A flavoprotein monooxygenase that inactivates a broad range of rifamycin antibiotics. Our findings describe a mechanism of rifamycin inactivation initiated by monooxygenation of the 2-position of the naphthyl group, which subsequently results in ring opening and linearization of the antibiotic. The result is an antibiotic that no longer adopts the basket-like structure essential for binding to the RNA exit tunnel of the target RpoB, thereby providing the molecular logic of resistance. This unique mechanism of enzymatic inactivation underpins the broad spectrum of rifamycin resistance mediated by Rox enzymes and presents a new antibiotic resistance mechanism not yet seen in microbial antibiotic detoxification.
Sigma-Aldrich
Rosetta-gami B(DE3)pLysS Competent Cells - Novagen, Rosetta-gami B strains combines the key features of BL21, Origami, and Rosetta to enhances the expression of eukaryotic proteins. T7 lysozyme expression suppresses basal T7 expression.
Pricing and availability is not currently available.