Merck
  • Functional enhancer elements drive subclass-selective expression from mouse to primate neocortex.
Functional enhancer elements drive subclass-selective expression from mouse to primate neocortex.
Cell reports (2021-04-01)
John K Mich, Lucas T Graybuck, Erik E Hess, Joseph T Mahoney, Yoshiko Kojima, Yi Ding, Saroja Somasundaram, Jeremy A Miller, Brian E Kalmbach, Cristina Radaelli, Bryan B Gore, Natalie Weed, Victoria Omstead, Yemeserach Bishaw, Nadiya V Shapovalova, Refugio A Martinez, Olivia Fong, Shenqin Yao, Marty Mortrud, Peter Chong, Luke Loftus, Darren Bertagnolli, Jeff Goldy, Tamara Casper, Nick Dee, Ximena Opitz-Araya, Ali Cetin, Kimberly A Smith, Ryder P Gwinn, Charles Cobbs, Andrew L Ko, Jeffrey G Ojemann, C Dirk Keene, Daniel L Silbergeld, Susan M Sunkin, Viviana Gradinaru, Gregory D Horwitz, Hongkui Zeng, Bosiljka Tasic, Ed S Lein, Jonathan T Ting, Boaz P Levi
ABSTRACT
Viral genetic tools that target specific brain cell types could transform basic neuroscience and targeted gene therapy. Here, we use comparative open chromatin analysis to identify thousands of human-neocortical-subclass-specific putative enhancers from across the genome to control gene expression in adeno-associated virus (AAV) vectors. The cellular specificity of reporter expression from enhancer-AAVs is established by molecular profiling after systemic AAV delivery in mouse. Over 30% of enhancer-AAVs produce specific expression in the targeted subclass, including both excitatory and inhibitory subclasses. We present a collection of Parvalbumin (PVALB) enhancer-AAVs that show highly enriched expression not only in cortical PVALB cells but also in some subcortical PVALB populations. Five vectors maintain PVALB-enriched expression in primate neocortex. These results demonstrate how genome-wide open chromatin data mining and cross-species AAV validation can be used to create the next generation of non-species-restricted viral genetic tools.
Sigma-Aldrich
Insulin solution from bovine pancreas, 10 mg/mL insulin in 25  mM HEPES, pH 8.2, BioReagent, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
Trypsin inhibitor from chicken egg white, Type II-O, Partially purified ovomucoid, containing ovoinhibitor
Sigma-Aldrich
Milli-Mark® Anti-NeuN-PE Antibody, clone A60, clone A60, Milli-Mark®, from mouse
Sigma-Aldrich
Anti-GABA Antibody, serum, Chemicon®
Sigma-Aldrich
Minimum Essential Medium Eagle, With Hanks′ salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining