Merck
  • Autophagy modulates endothelial junctions to restrain neutrophil diapedesis during inflammation.
Autophagy modulates endothelial junctions to restrain neutrophil diapedesis during inflammation.
Immunity (2021-08-08)
Natalia Reglero-Real, Lorena Pérez-Gutiérrez, Azumi Yoshimura, Loïc Rolas, José Garrido-Mesa, Anna Barkaway, Catherine Pickworth, Rebeca S Saleeb, Maria Gonzalez-Nuñez, Shani N Austin-Williams, Dianne Cooper, Laura Vázquez-Martínez, Tao Fu, Giulia De Rossi, Matthew Golding, Mathieu-Benoit Voisin, Chantal M Boulanger, Yoshiaki Kubota, William A Muller, Sharon A Tooze, Thomas D Nightingale, Lucy Collinson, Mauro Perretti, Ezra Aksoy, Sussan Nourshargh
ABSTRACT
The migration of neutrophils from the blood circulation to sites of infection or injury is a key immune response and requires the breaching of endothelial cells (ECs) that line the inner aspect of blood vessels. Unregulated neutrophil transendothelial cell migration (TEM) is pathogenic, but the molecular basis of its physiological termination remains unknown. Here, we demonstrated that ECs of venules in inflamed tissues exhibited a robust autophagic response that was aligned temporally with the peak of neutrophil trafficking and was strictly localized to EC contacts. Genetic ablation of EC autophagy led to excessive neutrophil TEM and uncontrolled leukocyte migration in murine inflammatory models, while pharmacological induction of autophagy suppressed neutrophil infiltration into tissues. Mechanistically, autophagy regulated the remodeling of EC junctions and expression of key EC adhesion molecules, facilitating their intracellular trafficking and degradation. Collectively, we have identified autophagy as a modulator of EC leukocyte trafficking machinery aimed at terminating physiological inflammation.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Zymosan A from Saccharomyces cerevisiae, for inducing inflamatory response
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Endothelial cell growth supplement from bovine neural tissue, ECGS, suitable for cell culture
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Tyrode′s Solution, Acidic, liquid, sterile-filtered, suitable for mouse embryo cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg
Sigma-Aldrich
Propidium iodide solution