Merck
  • COMMD4 functions with the histone H2A-H2B dimer for the timely repair of DNA double-strand breaks.
COMMD4 functions with the histone H2A-H2B dimer for the timely repair of DNA double-strand breaks.
Communications biology (2021-04-21)
Amila Suraweera, Neha S Gandhi, Sam Beard, Joshua T Burgess, Laura V Croft, Emma Bolderson, Ali Naqi, Nicholas W Ashton, Mark N Adams, Kienan I Savage, Shu-Dong Zhang, Kenneth J O'Byrne, Derek J Richard
ABSTRACT
Genomic stability is critical for normal cellular function and its deregulation is a universal hallmark of cancer. Here we outline a previously undescribed role of COMMD4 in maintaining genomic stability, by regulation of chromatin remodelling at sites of DNA double-strand breaks. At break-sites, COMMD4 binds to and protects histone H2B from monoubiquitination by RNF20/RNF40. DNA damage-induced phosphorylation of the H2A-H2B heterodimer disrupts the dimer allowing COMMD4 to preferentially bind H2A. Displacement of COMMD4 from H2B allows RNF20/40 to monoubiquitinate H2B and for remodelling of the break-site. Consistent with this critical function, COMMD4-deficient cells show excessive elongation of remodelled chromatin and failure of both non-homologous-end-joining and homologous recombination. We present peptide-mapping and mutagenesis data for the potential molecular mechanisms governing COMMD4-mediated chromatin regulation at DNA double-strand breaks.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-BRE1-A (RNF20) Antibody, clone 8A4.2, clone 8A4.2, from mouse
Sigma-Aldrich
Anti-53BP1 Antibody, clone BP13, clone BP13, Chemicon®, from mouse
Sigma-Aldrich
Rabbit anti-MDC1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
Sigma-Aldrich
Anti-p53 antibody, Mouse monoclonal, clone DO-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Histone H3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Supelco
6α-Naloxol solution, 1.0 mg/mL in methanol, certified reference material, Cerilliant®
Sigma-Aldrich
Normal Sheep IgG, Normal Sheep IgG Polyclonal Antibody control validated for use in Western Blotting.