Merck
  • Interleukin-34-CSF1R Signaling Axis Promotes Epithelial Cell Transformation and Breast Tumorigenesis.
Interleukin-34-CSF1R Signaling Axis Promotes Epithelial Cell Transformation and Breast Tumorigenesis.
International journal of molecular sciences (2021-04-04)
Muna Poudel, Garam Kim, Poshan Yugal Bhattarai, Jin-Young Kim, Hong Seok Choi
ABSTRACT
IL-34 has been recently identified as a ligand for CSF1R that regulates various cellular processes including cell proliferation, survival, and differentiation. Although the binding of IL-34 to CSF1R modulates several cancer-driving signaling pathways, little is known about the role of IL-34/CSF1R signaling in breast cancer. Herein, we report that IL-34 induces epithelial cell transformation and breast tumorigenesis through activation of MEK/ERK and JNK/c-Jun pathways. IL-34 increased the phosphorylation of MEK1/2, ERK1/2, JNK1/2, and c-Jun through CSF1R in mouse skin epidermal JB6 C141 cells and human breast cancer MCF7 cells. IL-34 enhanced c-Fos and c-Jun promoter activity, resulting in increased AP-1 transactivation activity in JB6 Cl41 and MCF7 cells. Moreover, PIN1 enhanced IL-34-induced phosphorylation of MEK1/2, ERK1/2, JNK1/2, and c-Jun in JB6 Cl41 and MCF7 cells. Inhibition of PIN1 using juglone prevented the IL-34-induced transformation of JB6 C141 cells. Similarly, silencing of PIN1 reduced the IL-34-induced tumorigenicity of MCF7 cells. Consistent with these results, the synergistic model showed that treatment with juglone suppressed the IL-34-induced growth of tumors formed by 4T1 cells in BALB/c mice. Our study demonstrates the role of IL-34-induced MEK/ERK and JNK/c-Jun cascades in breast cancer and highlights the regulatory role of PIN1 in IL-34-induced breast tumorigenesis.
Product Description
Juglone, phyproof® Reference Substance
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
5-Hydroxy-1,4-naphthoquinone, 97%
Sigma-Aldrich
PD 98059, PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.
Sigma-Aldrich
JNK Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC₅₀ = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).
Roche
Cell Proliferation ELISA, BrdU (colorimetric), sufficient for ≤1,000 tests