Merck
  • Identification of BBOX1 as a Therapeutic Target in Triple-Negative Breast Cancer.
Identification of BBOX1 as a Therapeutic Target in Triple-Negative Breast Cancer.
Cancer discovery (2020-07-22)
Chengheng Liao, Yang Zhang, Cheng Fan, Laura E Herring, Juan Liu, Jason W Locasale, Mamoru Takada, Jin Zhou, Giada Zurlo, Lianxin Hu, Jeremy M Simon, Travis S Ptacek, Victor G Andrianov, Einars Loza, Yan Peng, Huanghe Yang, Charles M Perou, Qing Zhang
ABSTRACT
Triple-negative breast cancer (TNBC) is an aggressive and highly lethal disease. Because of its heterogeneity and lack of hormone receptors or HER2 expression, targeted therapy is limited. Here, by performing a functional siRNA screening for 2-OG-dependent enzymes, we identified gamma-butyrobetaine hydroxylase 1 (BBOX1) as an essential gene for TNBC tumorigenesis. BBOX1 depletion inhibits TNBC cell growth while not affecting normal breast cells. Mechanistically, BBOX1 binds with the calcium channel inositol-1,4,5-trisphosphate receptor type 3 (IP3R3) in an enzymatic-dependent manner and prevents its ubiquitination and proteasomal degradation. BBOX1 depletion suppresses IP3R3-mediated endoplasmic reticulum calcium release, therefore impairing calcium-dependent energy-generating processes including mitochondrial respiration and mTORC1-mediated glycolysis, which leads to apoptosis and impaired cell-cycle progression in TNBC cells. Therapeutically, genetic depletion or pharmacologic inhibition of BBOX1 inhibits TNBC tumor growth in vitro and in vivo. Our study highlights the importance of targeting the previously uncharacterized BBOX1-IP3R3-calcium oncogenic signaling axis in TNBC. SIGNIFICANCE: We provide evidence from unbiased screens that BBOX1 is a potential therapeutic target in TNBC and that genetic knockdown or pharmacologic inhibition of BBOX1 leads to decreased TNBC cell fitness. This study lays the foundation for developing effective BBOX1 inhibitors for treatment of this lethal disease.This article is highlighted in the In This Issue feature, p. 1611.
Sigma-Aldrich
Monoclonal Anti-BBOX1 antibody produced in mouse, clone 6H3, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
DMOG, ≥98% (HPLC)
Supelco
EPA 8260A Surrogate Standards Mix, certified reference material, 2000 μg/mL each component in methanol
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human BBOX1