Merck
  • Performance of a novel in vitro assay for skin sensitization based on activation of T lymphocytes.
Performance of a novel in vitro assay for skin sensitization based on activation of T lymphocytes.
ALTEX (2020-04-24)
Fenxia Hou, Caihong Xing, Bin Li, Juan Cheng, Wei Chen
ABSTRACT
There are currently no skin sensitization assays based on T cell activation. We built a novel in vitro model to assess T cell activation and test its performance to discriminate skin sensitizers from non-sensitizers using 52 reference chemicals. Jurkat Clone E6-1 human T lymphocytes were exposed to a series of concentrations of test substances for 24 hours and CD69 expression was measured as a marker of early T cell activation with flow cytometry. Most tested sensitizers induced increased relative fluorescence intensity (RFI) of CD69 on the T cells, which was linearly correlated with the concentrations tested, indicating a statistically significant causal link between sensitizer concentration and increase of CD69 expression. CD69 RFI ≥ 1.5 was determined as the positive criterion for skin sensitizer classification. The sensitivity (79.4%), specificity (88.9%) and accuracy (82.7%) of the model for the 52 tested reference chemicals showed a good predictivity for skin sensitizers. The results were reproduced in at least two repeats; and the concurrent positive control, 2 mg/mL 2, 4-dinitrochlorobenzene, was found positive in all 25 independent runs conducted, indicating in-house reproducibility. The EC1.5 value (i.e., the concentration at which a test chemical induces a CD69 RFI of 1.5) may be used to assess skin sensitization potency of a chemical. This work may contribute to the development of an in vitro assay for skin sensitization based on the activation of T cells.
Sigma-Aldrich
Imidazolidinyl urea
Sigma-Aldrich
Nickel(II) sulfate hexa-/ heptahydrate, DIN 50970 H, ≥20.6% Ni and Co basis
Sigma-Aldrich
Phenyl benzoate, 99%
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
Globulins Cohn fraction II, III human, lyophilized powder
Sigma-Aldrich
2-Methyl-3-(p-isopropylphenyl)propionaldehyde, ≥95%, FCC, FG
Sigma-Aldrich
Nickel(II) sulfate, anhydrous, 99.99% trace metals basis