Merck
  • Potent and Selective Human Prostaglandin F (FP) Receptor Antagonist (BAY-6672) for the Treatment of Idiopathic Pulmonary Fibrosis (IPF).
Potent and Selective Human Prostaglandin F (FP) Receptor Antagonist (BAY-6672) for the Treatment of Idiopathic Pulmonary Fibrosis (IPF).
Journal of medicinal chemistry (2020-09-25)
Hartmut Beck, Tobias Thaler, Daniel Meibom, Mark Meininghaus, Hannah Jörißen, Lisa Dietz, Carsten Terjung, Michaela Bairlein, Clemens-Jeremias von Bühler, Sonja Anlauf, Chantal Fürstner, Timo Stellfeld, Dirk Schneider, Kersten M Gericke, Thomas Buyck, Kai Lovis, Uwe Münster, Johanna Anlahr, Elisabeth Kersten, Guillaume Levilain, Virginia Marossek, Raimund Kast
ABSTRACT
Idiopathic pulmonary fibrosis (IPF) is a rare and devastating chronic lung disease of unknown etiology. Despite the approved treatment options nintedanib and pirfenidone, the medical need for a safe and well-tolerated antifibrotic treatment of IPF remains high. The human prostaglandin F receptor (hFP-R) is widely expressed in the lung tissue and constitutes an attractive target for the treatment of fibrotic lung diseases. Herein, we present our research toward novel quinoline-based hFP-R antagonists, including synthesis and detailed structure-activity relationship (SAR). Starting from a high-throughput screening (HTS) hit of our corporate compound library, multiple parameter improvements-including increase of the relative oral bioavailability Frel from 3 to ≥100%-led to a highly potent and selective hFP-R antagonist with complete oral absorption from suspension. BAY-6672 (46) represents-to the best of our knowledge-the first reported FP-R antagonist to demonstrate in vivo efficacy in a preclinical animal model of lung fibrosis, thus paving the way for a new treatment option in IPF.
Sigma-Aldrich
BAY-6672 hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
3-Chloroperbenzoic acid, ≤77%