Merck
  • Quantification of citrullinated histones: Development of an improved assay to reliably quantify nucleosomal H3Cit in human plasma.
Quantification of citrullinated histones: Development of an improved assay to reliably quantify nucleosomal H3Cit in human plasma.
Journal of thrombosis and haemostasis : JTH (2020-07-13)
Charlotte Thålin, Katherina Aguilera, Nathan W Hall, Matthew R Marunde, Jonathan M Burg, Axel Rosell, Maud Daleskog, Maja Månsson, Yohei Hisada, Matthew J Meiners, Zu-Wen Sun, Matthew F Whelihan, Marcus A Cheek, Sarah A Howard, Shruti Saxena-Beem, Denis F Noubouossie, Nigel S Key, Saira Z Sheikh, Michael-Christopher Keogh, Martis W Cowles, Staffan Lundström, Nigel Mackman, Håkan Wallén, Andrea L Johnstone
ABSTRACT
Recent data propose a diagnostic and prognostic capacity for citrullinated histone H3 (H3Cit), a marker of neutrophil extracellular traps (NETs), in pathologic conditions such as cancer and thrombosis. However, current research is hampered by lack of standardized assays. We aimed to develop an assay to reliably quantify nucleosomal H3Cit in human plasma. We assessed the common practice of in vitro enzymatically modified histone H3 as calibration standards and the specificity of available intrapeptidyl citrulline antibodies. Based on our findings, we developed and validated a novel assay to quantify nucleosomal H3Cit in human plasma. We show that enzymatically citrullinated H3 proteins are compromised by high enzyme-dependent lot variability as well as instability in plasma. We furthermore demonstrate that the majority of commercially available antibodies against intrapeptidyl citrulline display poor specificity for their reported target when tested against a panel of semi-synthetic nucleosomes containing distinct histone H3 citrullinations. Finally, we present a novel assay utilizing highly specific monoclonal antibodies and semi-synthetic nucleosomes containing citrulline in place of arginine at histone H3, arginine residues 2, 8, and 17 (H3R2,8,17Cit) as calibration standards. Rigorous validation of this assay shows its capacity to accurately and reliably quantify nucleosomal H3Cit levels in human plasma with clear elevations in cancer patients compared to healthy individuals. Our novel approach using defined nucleosome controls enables reliable quantification of H3Cit in human plasma. This assay will be broadly applicable to study the role of histone citrullination in disease and its utility as a biomarker.
Sigma-Aldrich
Intralipid, 20%, emulsion
Sigma-Aldrich
Anti-Modified Citrulline Antibody, clone C4, clone C4, from human(Recombinant)
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
Anti-Citrulline Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-peptidyl-citrulline, clone F95 Antibody, clone F95, from mouse
Sigma-Aldrich
Anti-Citrulline Antibody, Upstate®, from rabbit