Merck
  • Attached stratified mucus separates bacteria from the epithelial cells in COPD lungs.
Attached stratified mucus separates bacteria from the epithelial cells in COPD lungs.
JCI insight (2018-09-07)
Joan Antoni Fernández-Blanco, Dalia Fakih, Liisa Arike, Ana M Rodríguez-Piñeiro, Beatriz Martínez-Abad, Elin Skansebo, Sonya Jackson, James Root, Dave Singh, Christopher McCrae, Christopher M Evans, Annika Åstrand, Anna Ermund, Gunnar C Hansson
ABSTRACT
The respiratory tract is normally kept essentially free of bacteria by cilia-mediated mucus transport, but in chronic obstructive pulmonary disease (COPD) and cystic fibrosis (CF), bacteria and mucus accumulates instead. To address the mechanisms behind the mucus accumulation, the proteome of bronchoalveolar lavages from COPD patients and mucus collected in an elastase-induced mouse model of COPD was analyzed, revealing similarities with each other and with the protein content in colonic mucus. Moreover, stratified laminated sheets of mucus were observed in airways from patients with CF and COPD and in elastase-exposed mice. On the other hand, the mucus accumulation in the elastase model was reduced in Muc5b-KO mice. While mucus plugs were removed from airways by washing with hypertonic saline in the elastase model, mucus remained adherent to epithelial cells. Bacteria were trapped on this mucus, whereas, in non-elastase-treated mice, bacteria were found on the epithelial cells. We propose that the adherence of mucus to epithelial cells observed in CF, COPD, and the elastase-induced mouse model of COPD separates bacteria from the surface cells and, thus, protects the respiratory epithelium.
Product Description
Millipore
MILLIPLEX® Mouse Cytokine/Chemokine Magnetic Bead Panel II, MECY2MAG-73K, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Millipore
MILLIPLEX® Mouse Cytokine/Chemokine Magnetic Bead Panel - Premixed 25 Plex - Space Saver (Bulk) Packaging, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.
Sigma-Aldrich
Monoclonal Anti-Mucin Gastric antibody produced in mouse, clone 45M1, ascites fluid