Merck
  • Myelination of parvalbumin interneurons shapes the function of cortical sensory inhibitory circuits.
Myelination of parvalbumin interneurons shapes the function of cortical sensory inhibitory circuits.
Nature communications (2020-10-15)
Najate Benamer, Marie Vidal, Maddalena Balia, María Cecilia Angulo
ABSTRACT
Myelination of projection neurons by oligodendrocytes is key to optimize action potential conduction over long distances. However, a large fraction of myelin enwraps the axons of parvalbumin-positive fast-spiking interneurons (FSI), exclusively involved in local cortical circuits. Whether FSI myelination contributes to the fine-tuning of intracortical networks is unknown. Here we demonstrate that FSI myelination is required for the establishment and maintenance of the powerful FSI-mediated feedforward inhibition of cortical sensory circuits. The disruption of GABAergic synaptic signaling of oligodendrocyte precursor cells prior to myelination onset resulted in severe FSI myelination defects characterized by longer internodes and nodes, aberrant myelination of branch points and proximal axon malformation. Consequently, high-frequency FSI discharges as well as FSI-dependent postsynaptic latencies and strengths of excitatory neurons were reduced. These dysfunctions generated a strong excitation-inhibition imbalance that correlated with whisker-dependent texture discrimination impairments. FSI myelination is therefore critical for the function of mature cortical inhibitory circuits.
Sigma-Aldrich
Anti-Ankyrin Antibody, clone Ank016, clone Ank016, Chemicon®, from mouse
Supelco
Naltrexone-3-β-D-glucuronide solution, 100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse