Merck
  • CYRI-A limits invasive migration through macropinosome formation and integrin uptake regulation.
CYRI-A limits invasive migration through macropinosome formation and integrin uptake regulation.
The Journal of cell biology (2021-06-25)
Anh Hoang Le, Tamas Yelland, Nikki R Paul, Loic Fort, Savvas Nikolaou, Shehab Ismail, Laura M Machesky
ABSTRACT
The Scar/WAVE complex drives actin nucleation during cell migration. Interestingly, the same complex is important in forming membrane ruffles during macropinocytosis, a process mediating nutrient uptake and membrane receptor trafficking. Mammalian CYRI-B is a recently described negative regulator of the Scar/WAVE complex by RAC1 sequestration, but its other paralogue, CYRI-A, has not been characterized. Here, we implicate CYRI-A as a key regulator of macropinosome formation and integrin internalization. We find that CYRI-A is transiently recruited to nascent macropinosomes, dependent on PI3K and RAC1 activity. CYRI-A recruitment precedes RAB5A recruitment but follows sharply after RAC1 and actin signaling, consistent with it being a local inhibitor of actin polymerization. Depletion of both CYRI-A and -B results in enhanced surface expression of the α5β1 integrin via reduced internalization. CYRI depletion enhanced migration, invasion, and anchorage-independent growth in 3D. Thus, CYRI-A is a dynamic regulator of macropinocytosis, functioning together with CYRI-B to regulate integrin trafficking.
Product Description
GSTrap 4B, Cytiva 28-4017-47, pack of 1 × 5 mL
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Integrin alpha5 (Preservative Free) Antibody, clone SNAKA51, clone SNAKA51, from mouse, purified by using protein G
Sigma-Aldrich
Anti-FAM49B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
Sigma-Aldrich
Anti-MMP-14 Antibody, catalytic domain, clone LEM-2/63.1, clone LEM-2/63.1, Chemicon®, from mouse
MBPTrap HP, Cytiva 28-9187-79, pack of 1 × 5 mL