Merck
  • Direct RNA sequencing reveals m6A modifications on adenovirus RNA are necessary for efficient splicing.
Direct RNA sequencing reveals m6A modifications on adenovirus RNA are necessary for efficient splicing.
Nature communications (2020-11-28)
Alexander M Price, Katharina E Hayer, Alexa B R McIntyre, Nandan S Gokhale, Jonathan S Abebe, Ashley N Della Fera, Christopher E Mason, Stacy M Horner, Angus C Wilson, Daniel P Depledge, Matthew D Weitzman
ABSTRACT
Adenovirus is a nuclear replicating DNA virus reliant on host RNA processing machinery. Processing and metabolism of cellular RNAs can be regulated by METTL3, which catalyzes the addition of N6-methyladenosine (m6A) to mRNAs. While m6A-modified adenoviral RNAs have been previously detected, the location and function of this mark within the infectious cycle is unknown. Since the complex adenovirus transcriptome includes overlapping spliced units that would impede accurate m6A mapping using short-read sequencing, here we profile m6A within the adenovirus transcriptome using a combination of meRIP-seq and direct RNA long-read sequencing to yield both nucleotide and transcript-resolved m6A detection. Although both early and late viral transcripts contain m6A, depletion of m6A writer METTL3 specifically impacts viral late transcripts by reducing their splicing efficiency. These data showcase a new technique for m6A discovery within individual transcripts at nucleotide resolution, and highlight the role of m6A in regulating splicing of a viral pathogen.
Sigma-Aldrich
Anti-METTL14 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
4-Thiouridine, ≥98%
Sigma-Aldrich
Anti-ALKBH5 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-YTHDF3 (ab1) antibody produced in rabbit, affinity isolated antibody
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
3-Deazaadenosine