Merck
  • Synthesis and pharmacological evaluation of novel conformationally constrained homologues of glutamic acid.
Synthesis and pharmacological evaluation of novel conformationally constrained homologues of glutamic acid.
European journal of medicinal chemistry (2007-03-10)
Paola Conti, Antonio Caligiuri, Andrea Pinto, Gabriella Roda, Lucia Tamborini, Birgitte Nielsen, Ulf Madsen, Karla Frydenvang, Alessio Colombo, Carlo De Micheli
ABSTRACT
Twelve novel conformationally constrained homologues of glutamic acid have been synthesized and pharmacologically characterized at ionotropic glutamate receptors (iGluRs). Synthesis of the target compounds involved 1,3-dipolar cycloaddition of nitrile oxides to suitable dipolarophiles. The structure to the compounds has been assigned by (1)H NMR and, in the case of derivatives (+/-)-4a, (+/-)-4b, (+/-)-5a, and (+/-)-5b, by means of an X-ray crystallographic analysis carried out on intermediate (+/-)-12a. The synthesized amino acids were found to be without affinity (K(i)/IC(50)>100microM) for iGluRs with the exception of compounds (+/-)-4b and (+/-)-5b, which showed a modest affinity for NMDA receptors (K(i)=34 and 13microM, respectively). The results indicate that the increased conformational constraints introduced by the cyclopropane ring and the spiro-attached proline ring are both detrimental to the pharmacological activity.
Sigma-Aldrich
D(−)-2-Amino-5-phosphonopentanoic acid, NMDA receptor antagonist
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
5 g
Please contact Customer Service for Availability
€187.00
25 g
Please contact Customer Service for Availability
€657.00
To order products, please contact your local dealer.