Merck
  • Hypothalamic Contribution to Pituitary Functions Is Recapitulated In Vitro Using 3D-Cultured Human iPS Cells.
Hypothalamic Contribution to Pituitary Functions Is Recapitulated In Vitro Using 3D-Cultured Human iPS Cells.
Cell reports (2020-01-09)
Takatoshi Kasai, Hidetaka Suga, Mayu Sakakibara, Chikafumi Ozone, Ryusaku Matsumoto, Mayuko Kano, Kazuki Mitsumoto, Koichiro Ogawa, Yu Kodani, Hiroshi Nagasaki, Naoko Inoshita, Mariko Sugiyama, Takeshi Onoue, Taku Tsunekawa, Yoshihiro Ito, Hiroshi Takagi, Daisuke Hagiwara, Shintaro Iwama, Motomitsu Goto, Ryoichi Banno, Jun Takahashi, Hiroshi Arima
ABSTRACT
The pituitary is a major hormone center that secretes systemic hormones responding to hypothalamus-derived-releasing hormones. Previously, we reported the independent pituitary induction and hypothalamic differentiation of human embryonic stem cells (ESCs). Here, a functional hypothalamic-pituitary unit is generated using human induced pluripotent stem (iPS) cells in vitro. The adrenocorticotropic hormone (ACTH) secretion capacity of the induced pituitary reached a comparable level to that of adult mouse pituitary because of the simultaneous maturation with hypothalamic neurons within the same aggregates. Corticotropin-releasing hormone (CRH) from the hypothalamic area regulates ACTH cells similarly to our hypothalamic-pituitary axis. Our induced hypothalamic-pituitary units respond to environmental hypoglycemic condition in vitro, which mimics a life-threatening situation in vivo, through the CRH-ACTH pathway, and succeed in increasing ACTH secretion. Thus, we generated powerful hybrid organoids by recapitulating hypothalamic-pituitary development, showing autonomous maturation on the basis of interactions between developing tissues.
Sigma-Aldrich
NBI 27914, ≥98% (HPLC)
Sigma-Aldrich
Antalarmin hydrochloride, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Gold antibody produced in goat, affinity isolated antibody, aqueous glycerol suspension, 10 nm (colloidal gold)
Sigma-Aldrich
Monoclonal Anti-Cytokeratin, pan (Mixture) antibody produced in mouse, clone C-11+PCK-26+CY-90+KS-1A3+M20+A53-B/A2, ascites fluid
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, γ-irradiated, Globulin Free, BioXtra, suitable for cell culture
Sigma-Aldrich
Anti-MCH antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Oxytocin Antibody, clone 4G11, ascites fluid, clone 4G11, Chemicon®
Sigma-Aldrich
1-Thioglycerol, liquid, BioReagent, suitable for cell culture, ≥97% (titration)
Sigma-Aldrich
Earle′s Balanced Salts, With sodium bicarbonate, without phenol red, liquid, sterile-filtered, suitable for cell culture