Merck
  • Novel serum peptide model revealed by MALDI-TOF-MS and its diagnostic value in early bladder cancer.
Novel serum peptide model revealed by MALDI-TOF-MS and its diagnostic value in early bladder cancer.
The International journal of biological markers (2020-07-24)
Dapeng Ding, Mingying Chen, Xiaoguang Xiao, Penglong Cao, Shijun Li
ABSTRACT
Bladder cancer is the ninth most common cancer worldwide and has high morbidity and mortality. We aimed to search for potential serum peptide biomarkers and establish a diagnostic model for early bladder cancer. A total of 67 bladder cancer patients and 64 healthy volunteers were randomly divided into a training set and testing set 1. There were 30 hematuria patients used as testing set 2. Matrix-assisted laser desorption/ionization time-of-flight mass spectrometry based on weak cation exchange magnetic beads was used to obtain and analyze the serum peptide profiles between bladder cancer patients and healthy volunteers in the training set. Serum peptide diagnostic model through a k-nearest neighbor algorithm, was established and validated, and significantly differentially expressed protein biomarkers were ultimately identified. We constructed a diagnostic model containing five peptides (m/z 1954.9, m/z 2081.0, m/z 3938.3, m/z 3946.5, and m/z 4268.8). In the training set, the area under the curve (AUC) value of the five-peptide model was 0.923, and the sensitivity and specificity was 93.75% and 96.77%, respectively. In testing set 1, the sensitivity and specificity was 91.43% and 90.91%, respectively, and the specificity of testing set 2 was 73.33%. For early-stage bladder cancer, the sensitivity and specificity was 92.31% and 93.75%, respectively; the sensitivity of early low-grade bladder cancer was 90.00%; and the AUC value was 0.944. The five-peptide diagnostic model established in this study had high sensitivity and specificity, especially in the diagnosis of early bladder cancer, and could differentiate between healthy volunteers and hematuria patients.
Sigma-Aldrich
ProteoMass ACTH Fragment 18-39 MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 2,464.1989 Da
Pricing and availability is not currently available.
Sigma-Aldrich
ProteoMass P14R MALDI-MS Standard, vial of 10 nmol
Pricing and availability is not currently available.
Sigma-Aldrich
ProteoMass Insulin MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 5,729.6087 Da
Pricing and availability is not currently available.