Merck
AUTACs: Cargo-Specific Degraders Using Selective Autophagy.
Molecular cell (2019-10-14)
Daiki Takahashi, Jun Moriyama, Tomoe Nakamura, Erika Miki, Eriko Takahashi, Ayami Sato, Takaaki Akaike, Kaori Itto-Nakama, Hirokazu Arimoto
ABSTRACT
Protein silencing represents an essential tool in biomedical research. Targeted protein degradation (TPD) strategies exemplified by PROTACs are rapidly emerging as modalities in drug discovery. However, the scope of current TPD techniques is limited because many intracellular materials are not substrates of proteasomal clearance. Here, we described a novel targeted-clearance strategy (autophagy-targeting chimera [AUTAC]) that contains a degradation tag (guanine derivatives) and a warhead to provide target specificity. As expected from the substrate scope of autophagy, AUTAC degraded fragmented mitochondria as well as proteins. Mitochondria-targeted AUTAC accelerated both the removal of dysfunctional fragmented mitochondria and the biogenesis of functionally normal mitochondria in patient-derived fibroblast cells. Cytoprotective effects against acute mitochondrial injuries were also seen. Canonical autophagy is viewed as a nonselective bulk decomposition system, and none of the available autophagy-inducing agents exhibit useful cargo selectivity. With its target specificity, AUTAC provides a new modality for research on autophagy-based drugs.
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Sigma-Aldrich
Anti-Ubiquitin Antibody, Lys63-Specific, clone Apu3, rabbit monoclonal, clone Apu3, from rabbit
Sigma-Aldrich
PYR-41, ≥98% (HPLC), powder
Sigma-Aldrich
N-Acetyl-S-(2-amino-9-(4-fluorobenzyl)-6-oxo-6,9-dihydro-1H-purin-8-yl)-L-cysteine, ≥95%
Sigma-Aldrich
FBnG-C3-PEG1-C3-NH2 hydrochloride, ≥95%
Sigma-Aldrich
FBnG-C3-PEG5-C3-NH2 hydrochloride, ≥95%