Merck
Biodegradable Metal-Organic Framework-Based Microrobots (MOFBOTs).
Advanced healthcare materials (2020-09-10)
Anastasia Terzopoulou, Xiaopu Wang, Xiang-Zhong Chen, Mario Palacios-Corella, Carlos Pujante, Javier Herrero-Martín, Xiao-Hua Qin, Jordi Sort, Andrew J deMello, Bradley J Nelson, Josep Puigmartí-Luis, Salvador Pané
ABSTRACT
Microrobots and metal-organic frameworks (MOFs) have been identified as promising carriers for drug delivery applications. While clinical applications of microrobots are limited by their low drug loading efficiencies and the poor degradability of the materials used for their fabrication, MOFs lack motility and targeted drug delivery capabilities. The combination of these two fields marks the beginning of a new era; MOF-based small-scale robots (MOFBOTs) for biomedical applications. Yet, biodegradability is a major hurdle in the field of micro- and nanoswimmers including small-scale robots. Here, a highly integrated MOFBOT that is able to realize magnetic locomotion, drug delivery, and selective degradation in cell cultures is reported for the first time. The MOF used in the investigations does not only allow a superior loading of chemotherapeutic drugs and their controlled release via a pH-responsive degradation but it also enables the controlled locomotion of enzymatically biodegradable gelatin-based helical microrobots under magnetic fields. The degradation of the integrated MOFBOT is observed after two weeks, when all its components fully degrade. Additionally, drug delivery studies performed in cancer cell cultures show reduced viability upon delivery of Doxorubicin within short time frames. This MOFBOT system opens new avenues for highly integrated fully biodegradable small-scale robots.
Sigma-Aldrich
Zinc acetate dihydrate, ACS reagent, ≥98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Phalloidin–Tetramethylrhodamine B isothiocyanate, sequence from Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
2-Methylimidazole, 99%
Supelco
Doxorubicin Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ACS reagent, ≥99.0%