Merck
  • Comparison of eight methods for the extraction of Bacillus atrophaeus spore DNA from eleven common interferents and a common swab.
Comparison of eight methods for the extraction of Bacillus atrophaeus spore DNA from eleven common interferents and a common swab.
PloS one (2011-08-06)
Helen L Rose, Caroline A Dewey, Morgan S Ely, Sarah L Willoughby, Tanya M Parsons, Victoria Cox, Phillippa M Spencer, Simon A Weller
ABSTRACT
Eight DNA extraction products or methods (Applied Biosystems PrepFiler Forensic DNA Extraction Kit; Bio-Rad Instagene Only, Bio-Rad Instagene & Spin Column Purification; EpiCentre MasterPure DNA & RNA Kit; FujiFilm QuickGene Mini80; Idaho Technologies 1-2-3 Q-Flow Kit; MoBio UltraClean Microbial DNA Isolation Kit; Sigma Extract-N-Amp Plant and Seed Kit) were adapted to facilitate extraction of DNA under BSL3 containment conditions. DNA was extracted from 12 common interferents or sample types, spiked with spores of Bacillus atropheaus. Resulting extracts were tested by real-time PCR. No one method was the best, in terms of DNA extraction, across all sample types. Statistical analysis indicated that the PrepFiler method was the best method from six dry powders (baking, biological washing, milk, plain flour, filler and talcum) and one solid (Underarm deodorant), the UltraClean method was the best from four liquids (aftershave, cola, nutrient broth, vinegar), and the MasterPure method was the best from the swab sample type. The best overall method, in terms of DNA extraction, across all sample types evaluated was the UltraClean method.
Sigma-Aldrich
JumpStart Taq DNA Polymerase, with MgCl2
Sigma-Aldrich
JumpStart Taq DNA Polymerase, without MgCl2
Sigma-Aldrich
Extract-N-Amp Seed PCR Kit, sufficient for 100 extractions, sufficient for 100 amplifications