Merck
  • Native Bovine Hydroxyapatite Powder, Demineralised Bone Matrix Powder, and Purified Bone Collagen Membranes Are Efficient in Repair of Critical-Sized Rat Calvarial Defects.
Native Bovine Hydroxyapatite Powder, Demineralised Bone Matrix Powder, and Purified Bone Collagen Membranes Are Efficient in Repair of Critical-Sized Rat Calvarial Defects.
Materials (Basel, Switzerland) (2020-08-06)
Alexey Veremeev, Roman Bolgarin, Vladimir Nesterenko, Alexander Andreev-Andrievskiy, Anton Kutikhin
ABSTRACT
Here we evaluated the efficacy of bone repair using various native bovine biomaterials (refined hydroxyapatite (HA), demineralised bone matrix (DBM), and purified bone collagen (COLL)) as compared with commercially available bone mineral and bone autografts. We employed a conventional critical-sized (8 mm diameter) rat calvarial defect model (6-month-old male Sprague-Dawley rats, n = 72 in total). The artificial defect was repaired using HA, DBM, COLL, commercially available bone mineral powder, bone calvarial autograft, or remained unfilled (n = 12 animals per group). Rats were euthanised 4 or 12 weeks postimplantation (n = 6 per time point) with the subsequent examination to assess the extent, volume, area, and mineral density of the repaired tissue by means of microcomputed tomography and hematoxylin and eosin staining. Bovine HA and DBM powder exhibited excellent repair capability similar to the autografts and commercially available bone mineral powder while COLL showed higher bone repair rate. We suggest that HA and DBM powder obtained from bovine bone tissue can be equally applied for the repair of bone defects and demonstrate sufficient potential to be implemented into clinical studies.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
IKA® sieves for MF 10 basic microfine grinder drive, MF 0.25, 0.25mm, 1/cs
IKA® sieves for MF 10 basic microfine grinder drive, MF 1.0, 1.0mm, 1/cs
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium chloride, ACS reagent, ≥99.0%
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, BioXtra