Merck
  • RPS25 is essential for translation initiation by the Dicistroviridae and hepatitis C viral IRESs.
RPS25 is essential for translation initiation by the Dicistroviridae and hepatitis C viral IRESs.
Genes & development (2009-12-03)
Dori M Landry, Marla I Hertz, Sunnie R Thompson
ABSTRACT
Most eukaryotic mRNAs are translated using a cap-dependent mechanism of translation. However, approximately 10% of mammalian mRNAs initiate translation using a cap-independent mechanism that is not well understood. These mRNAs contain an internal ribosome entry site (IRES) located in the 5' untranslated region. The cricket paralysis virus (CrPV) intergenic region IRES (IGR IRES) functions in yeast, mammals, and plants, and does not require any translation initiation factors. We used yeast genetics to understand how ribosomes are recruited directly to the mRNA by an IRES. We found that Rps25p has an essential role in CrPV IGR IRES activity in yeast and mammalian cells but not in cap-dependent translation. Purified 40S ribosomal subunits lacking Rps25 are unable to bind to the IGR IRES in vitro. The hepatitis C virus (HCV) IRES also requires Rps25, demonstrating the function of Rps25 is conserved across IRES types. Yeast strains lacking Rps25 exhibit only slight defects in global translation, readthrough, ribosome biogenesis, and programmed ribosomal frameshifting. This work is the first demonstration of a ribosomal protein that is specifically required for IRES-mediated translation initiation. Our findings provide us with the beginnings of a model for the molecular interactions of an IRES with the ribosome.
Product Description
Whatman® nitrocellulose membrane filters, plain white, pore size 1 μm, diam. 25 mm
Whatman® nitrocellulose membrane filters, plain white, pore size 3 μm, diam. 25 mm
Whatman® nitrocellulose membrane filters, plain white, pore size 0.1 μm, diam. 47 mm
Whatman® mixed cellulose ester membrane filters, gridded white (without pad), pore size 0.45 μm, diam. 47 mm, sterile
Whatman® nitrocellulose membrane filters, plain white, pore size 1 μm, diam. 47 mm
Whatman® mixed cellulose ester membrane filters, (white/black grid), pore size 0.2 μm, diam. 47 mm, sterile
Whatman® nitrocellulose membrane filters, pore size 0.1 μm, plain white, diam. 25 mm
Whatman® Mixed Cellulose Ester filters white gridded, pore size 0.45 μm, diam. 47 mm, sterile- pack of 100 ea, gridded white, pore size 0.45 μm, diam. 47 mm, sterile
Whatman® mixed cellulose ester membrane filters, gridded white, pore size 0.45 μm, diam. 47 mm, pack of 100 ea
Whatman® mixed cellulose ester membrane filters, gridded white, pore size 0.45 μm, diam. 47 mm, sterile
Whatman® nitrocellulose membrane filters, plain white, pore size 1.2 μm, diam. 50 mm
Whatman® nitrocellulose membrane filters, plain white, pore size 1.2 μm, size 142 mm
Whatman® mixed cellulose ester membrane filters, gridded black, pore size 0.45 μm, diam. 47 mm, sterile
Whatman® nitrocellulose membrane filters, NC10 Membrane Circles, diam. 50 mm, pore size 0.1 μm
Whatman® nitrocellulose membrane filters, NC10 Membrane Circles, diam. 47 mm, pore size 0.1 μm
Whatman® nitrocellulose membrane filters, pore size 12 μm, diam. 47 mm, pack of 100 ea
Whatman® nitrocellulose membrane filters, pore size 12 μm, diam. 50 mm, pack of 100 ea
Whatman® nitrocellulose membrane filters, plain white, pore size 3 μm, diam. 47 mm
Whatman® nitrocellulose membrane filters, plain white, pore size 0.2 μm, diam. 25 mm
Whatman® nitrocellulose membrane filters, NC45 Membrane Circles, diam. 47 mm, pore size 0.45 μm
Whatman® nitrocellulose membrane filters, pore size 8 μm, diam. 50 mm, pack of 100 ea
Whatman® nitrocellulose membrane filters, NC 45, pore size 0.45 μm
Whatman® nitrocellulose membrane filters, pore size 5 μm, diam. 25 mm, pack of 100 ea
Whatman® nitrocellulose membrane filters, NC45 Membrane Circles, diam. 47 mm, pore size 0.45 μm
Whatman® nitrocellulose membrane filters, plain white, pore size 0.8 μm, size 90 mm
Whatman® nitrocellulose membrane filters, plain white, pore size 0.45 μm, diam. 47 mm
Whatman® nitrocellulose membrane filters, pore size 5 μm, diam. 47 mm, pack of 100 ea
Whatman® nitrocellulose membrane filters, NC45 Membrane Circles, diam. 25 mm, pore size 0.45 μm
Whatman® nitrocellulose membrane filters, NC20 Membrane Circles, diam. 50 mm, pore size 0.2 μm
Whatman® nitrocellulose membrane filters, pore size 8 μm, diam. 47 mm, pack of 100 ea