Merck
  • Biodegradable cationic nanogels with tunable size, swelling and pKa for drug delivery.
Biodegradable cationic nanogels with tunable size, swelling and pKa for drug delivery.
International journal of pharmaceutics (2020-07-30)
D S Spencer, A B Shodeinde, D W Beckman, B C Luu, H R Hodges, N A Peppas
ABSTRACT
Cationic polymers have garnered significant interest for their utility in intracellular drug delivery and gene therapy. However, due to their associated toxicities, novel synthesis approaches must be explored to develop materials that are biocompatible. The novel library of nanoparticles synthesized in this study exhibit tunable hydrodynamic diameters, composition and pH-responsive properties as a function of synthesis parameters. In addition, differences in the responsiveness of these nanoparticles under different pH conditions affords greater control over intracellular drug release.
Sigma-Aldrich
(Trimethylsilyl)methacrylate, 98%
Sigma-Aldrich
2-(Trimethylsilyloxy)ethyl methacrylate, contains ≤100 ppm BHT as inhibitor, 96%
Sigma-Aldrich
tert-Butyl methacrylate, 98%, contains 200 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Benzyl methacrylate, 96%, contains monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
2-(Diisopropylamino)ethyl methacrylate, 97%, contains ~100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Bis(2-methacryloyl)oxyethyl disulfide, contains ≤6000 ppm hydroquinone as stabilizer
Sigma-Aldrich
Cyclohexyl methacrylate, ≥97%, contains ~60 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
6-(p-Toluidino)-2-naphthalenesulfonic acid sodium salt
Sigma-Aldrich
5-(N,N-Hexamethylene)amiloride