Merck
  • A Versatile Tiling Light Sheet Microscope for Imaging of Cleared Tissues.
A Versatile Tiling Light Sheet Microscope for Imaging of Cleared Tissues.
Cell reports (2020-11-05)
Yanlu Chen, Xiaoliang Li, Dongdong Zhang, Chunhui Wang, Ruili Feng, Xuzhao Li, Yao Wen, Hao Xu, Xinyi Shirley Zhang, Xiao Yang, Yongyi Chen, Yi Feng, Bo Zhou, Bi-Chang Chen, Kai Lei, Shang Cai, Jie-Min Jia, Liang Gao
ABSTRACT
We present a tiling light sheet microscope compatible with all tissue clearing methods for rapid multicolor 3D imaging of cleared tissues with micron-scale (4 × 4 × 10 μm3) to submicron-scale (0.3 × 0.3 × 1 μm3) spatial resolution. The resolving ability is improved to sub-100 nm (70 × 70 × 200 nm3) via tissue expansion. The microscope uses tiling light sheets to achieve higher spatial resolution and better optical sectioning ability than conventional light sheet microscopes. The illumination light is phase modulated to adjust the position and intensity profile of the light sheet based on the desired spatial resolution and imaging speed and to keep the microscope aligned. The ability of the microscope to align via phase modulation alone also ensures its accuracy for multicolor 3D imaging and makes the microscope reliable and easy to operate. Here we describe the working principle and design of the microscope. We demonstrate its utility by imaging various cleared tissues.
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Glycine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
N,N,N′,N′-Tetrakis(2-Hydroxypropyl)ethylenediamine, 98%
Sigma-Aldrich
Sodium acrylate, 97%
Sigma-Aldrich
Acrylamide, ≥99% (HPLC)
Sigma-Aldrich
Benzyl ether, 98%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Poly(ethylene glycol) methacrylate, average Mn 500, contains 900 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Benzyl benzoate, ReagentPlus®, ≥99.0%
Roche
Anti-Digoxigenin-POD, Fab fragments, from sheep
Sigma-Aldrich
tert-Butanol, ACS reagent, ≥99.0%