Merck
  • Ferguson analysis of protein electromigration during single-cell electrophoresis in an open microfluidic device.
Ferguson analysis of protein electromigration during single-cell electrophoresis in an open microfluidic device.
The Analyst (2020-04-30)
Kristine Y Tan, Amy E Herr
ABSTRACT
In an open microfluidic device, we investigate protein polyacrylamide gel electrophoresis (PAGE) separation performance on single-cell lysate. Single-cell protein electrophoresis is performed in a thin layer of polyacrylamide (PA) gel into which microwells are molded. Individual cells are isolated in these open microwells, then lysed on-chip with a dual lysis and electrophoresis sodium dodecyl sulfate (SDS) buffer. We scrutinize the effect of sieving gel composition on electromigration of protein targets, using a wide range of cellular protein standards (36 kDa to 289 kDa). We find that as PA concentration increases, protein electromigration deviates from the empirical log-linear relationship predicted between migration distance and molecular mass. We perform Ferguson analysis to calculate retardation coefficients and free solution mobilities of nine cellular protein standards and observe that the largest-molecular-mass protein, mTOR (289 kDa), does not behave as predicted by established linear-fit models for SDS-denatured proteins, indicating that mTOR is beyond the linear range of this assay. Lastly, we performed in-gel immunoprobing on the single-cell electrophoretic separations and observed that smaller pore-size gels (higher gel concentration) reduce protein diffusion out of the gel, which does not notably impact the measured immunoprobed protein expression. Compared to larger pore-size gels, the smaller pore-size gels lead to higher local concentrations of the target protein in each protein band, resulting in an increase in the signal-to-noise ratio (SNR) for each protein. Understanding the separation and immunoprobing performance at different gel concentrations improves assay design and optimization for target proteins.
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Dichlorodimethylsilane, ≥99.5%
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Anti-GAPDH, C-Terminal antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, 98%
Millipore
Hemin Chloride, Iron-containing compound of porcine origin that can induce hydrogen peroxide-dependent lipid peroxidation. Reported to increase the number and size of myotubes and the frequency of myotube contractions.
Sigma-Aldrich
Acrylamide/Bis-acrylamide, 30% solution, BioReagent, suitable for electrophoresis, 29:1
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)