Merck
  • Acetylation of Cytidine Residues Boosts HIV-1 Gene Expression by Increasing Viral RNA Stability.
Acetylation of Cytidine Residues Boosts HIV-1 Gene Expression by Increasing Viral RNA Stability.
Cell host & microbe (2020-06-14)
Kevin Tsai, Ananda Ayyappan Jaguva Vasudevan, Cecilia Martinez Campos, Ann Emery, Ronald Swanstrom, Bryan R Cullen
ABSTRACT
Epitranscriptomic RNA modifications, including methylation of adenine and cytidine residues, are now recognized as key regulators of both cellular and viral mRNA function. Moreover, acetylation of the N4 position of cytidine (ac4C) was recently reported to increase the translation and stability of cellular mRNAs. Here, we show that ac4C and N-acetyltransferase 10 (NAT10), the enzyme that adds ac4C to RNAs, have been subverted by human immunodeficiency virus 1 (HIV-1) to increase viral gene expression. HIV-1 transcripts are modified with ac4C at multiple discrete sites, and silent mutagenesis of these ac4C sites led to decreased HIV-1 gene expression. Similarly, loss of ac4C from viral transcripts due to depletion of NAT10 inhibited HIV-1 replication by reducing viral RNA stability. Interestingly, the NAT10 inhibitor remodelin could inhibit HIV-1 replication at concentrations that have no effect on cell viability, thus identifying ac4C addition as a potential target for antiviral drug development.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Sigma-Aldrich
Nevirapine
Sigma-Aldrich
4-Thiouridine, ≥98%
Sigma-Aldrich
Remodelin hydrobromide, ≥98% (HPLC)
Roche
Phytohemagglutinin-M (PHA-M), from Phaseolus vulgaris