Merck
  • Synthesis of isotopically labelled αCGRP8-37 and its lipidated analogue.
Synthesis of isotopically labelled αCGRP8-37 and its lipidated analogue.
Journal of labelled compounds & radiopharmaceuticals (2020-03-27)
Benjamin L Lu, Kerry M Loomes, Debbie L Hay, Paul W R Harris, Margaret A Brimble
ABSTRACT
α-Calcitonin gene related peptide (αCGRP) inhibitors are important medicinal targets due to their ability to produce antimigraine effects, thus, the discovery of long-acting αCGRP inhibitors is of significant interest. Herein we report the synthesis of an isotopically labelled version of the well-known CGRP receptor antagonist, αCGRP8-37 , as well as lipidated αCGRP8-37 with comparable antagonistic activity. These isotopically labelled peptides can be employed in assays to determine the metabolic stability of the lipidated αCGRP8-37 and compare this with the stability of known αCGRP8-37 .
Sigma-Aldrich
(2S)-6-Amino-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Sigma-Aldrich
2,2-Dimethoxy-2-phenylacetophenone, 99%
Sigma-Aldrich
2,2′-(Ethylenedioxy)diethanethiol, 95%