Merck
  • Pharmacoproteomics Identifies Kinase Pathways that Drive the Epithelial-Mesenchymal Transition and Drug Resistance in Hepatocellular Carcinoma.
Pharmacoproteomics Identifies Kinase Pathways that Drive the Epithelial-Mesenchymal Transition and Drug Resistance in Hepatocellular Carcinoma.
Cell systems (2020-08-07)
Martin Golkowski, Ho-Tak Lau, Marina Chan, Heidi Kenerson, Venkata Narayana Vidadala, Anna Shoemaker, Dustin J Maly, Raymond S Yeung, Taranjit S Gujral, Shao-En Ong
ABSTRACT
Hepatocellular carcinoma (HCC) is a complex and deadly disease lacking druggable genetic mutations. The limited efficacy of systemic treatments for advanced HCC implies that predictive biomarkers and drug targets are urgently needed. Most HCC drugs target protein kinases, indicating that kinase-dependent signaling networks drive HCC progression. To identify HCC signaling networks that determine responses to kinase inhibitors (KIs), we apply a pharmacoproteomics approach integrating kinome activity in 17 HCC cell lines with their responses to 299 KIs, resulting in a comprehensive dataset of pathway-based drug response signatures. By profiling patient HCC samples, we identify signatures of clinical HCC drug responses in individual tumors. Our analyses reveal kinase networks promoting the epithelial-mesenchymal transition (EMT) and drug resistance, including a FZD2-AXL-NUAK1/2 signaling module, whose inhibition reverses the EMT and sensitizes HCC cells to drugs. Our approach identifies cancer drug targets and molecular signatures of drug response for personalized oncology.
Sigma-Aldrich
MISSION® esiRNA, targeting human FZD2
Sigma-Aldrich
MISSION® esiRNA, targeting human NUAK1
Sigma-Aldrich
MISSION® esiRNA, targeting human NUAK2
Sigma-Aldrich
MISSION® esiRNA, targeting human AXL
Millipore
Inulin Discs, suitable for microbiology, Sterile filter paper discs impregnated with inulin
Sigma-Aldrich
MISSION® esiRNA, targeting human FYN
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
WZ4003, ≥98% (HPLC)
Sigma-Aldrich
Anti-Vimentin Antibody, clone V9, clone V9, Chemicon®, from mouse