Merck
  • Oxidation of multiple MiT/TFE transcription factors links oxidative stress to transcriptional control of autophagy and lysosome biogenesis.
Oxidation of multiple MiT/TFE transcription factors links oxidative stress to transcriptional control of autophagy and lysosome biogenesis.
Autophagy (2019-12-13)
Hongfeng Wang, Nana Wang, Delai Xu, Qilian Ma, Yang Chen, Shiqiang Xu, Qin Xia, Yan Zhang, Jochen H M Prehn, Guanghui Wang, Zheng Ying
ABSTRACT
Significant evidences indicate that reactive oxygen species (ROS) can induce macroautophagy/autophagy under both physiological and pathological conditions. Although the relationship between ROS and autophagy regulation has been well studied, the basic mechanism by which ROS affects autophagy and the biological role of this regulation are still not fully understood. In the present study we show that multiple MiT-TFE transcription factors including TFEB, TFE3 and MITF, which are master regulators of autophagy and lysosomal biogenesis, can be activated upon direct cysteine oxidation by ROS. Oxidation promotes the nuclear translocation of these MiT-TFE transcription factors by inhibiting the association of them with RRAG GTPases, which in turn leads to enhanced global gene expression level in autophagy-lysosome system. Our study highlights the role of oxidation of MiT-TFE transcription factors in ROS-linked autophagy, and provides novel mechanism that MiT-TFE transcription factors-mediated transcriptional control of autophagy may govern cell homeostasis in response to oxidative stress, a biological process tightly linked to human diseases including neurodegenerative diseases and cancer. Bafi A1: bafilomycin A1; EBSS: Earle's balanced salt solution; EGFP: enhanced green fluorescent protein; GAPDH: glyceraldehyde-3-phosphate dehydrogenase; MAP1LC3B/LC3B: microtubule associated protein 1 light chain 3 beta; MTORC1: mechanistic target of rapamycin kinase complex 1; ROS: reactive oxygen species; RPS6KB/p70S6K: ribosomal protein S6 kinase B; TFEB: transcription factor EB; WT: wild type.
Sigma-Aldrich
4-Nitroquinoline N-oxide, ≥98%
Sigma-Aldrich
Biotin-maleimide, ≥95% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Bafilomycin A1, Streptomyces griseus, Bafilomycin A1, CAS 88899-55-2, acts as a highly potent and specific inhibitor of vacuolar-type H+-ATPase (Ki = 500 pM). Blocks the fusion of autophagosome with lysosome.
Sigma-Aldrich
Leptomycin B solution from Streptomyces sp., ≥95% (HPLC), Supplied in methanol: water (7:3)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)