Merck
  • Metal oxides and annealed metals as alternatives to metal salts for fixed-ratio metal mixture ecotoxicity tests in soil.
Metal oxides and annealed metals as alternatives to metal salts for fixed-ratio metal mixture ecotoxicity tests in soil.
PloS one (2020-03-07)
Mathieu Renaud, Mark Cousins, Kobby Fred Awuah, Olukayode Jegede, Beverley Hale, José Paulo Sousa, Steven Douglas Siciliano
ABSTRACT
In soil metal ecotoxicology research, dosing is usually performed with metal salts, followed by leaching to remove excess salinity. This process also removes some metals, affecting metal mixture ratios as different metals are removed by leaching at different rates. Consequently, alternative dosing methods must be considered for fixed ratio metal mixture research. In this study three different metal mixture dosing methods (nitrate, oxide and annealed metal dosing) were examined for metal concentrations and toxicity. In the nitrate metal dosing method leaching reduced total metal retention and was affected by soil pH and cation exchange capacity (CEC). Acidic soils 3.22 (pH 3.4, CEC 8 meq/100g) and WTRS (pH 4.6, CEC 16 meq/100g) lost more than 75 and 64% of their total metals to leaching respectively while Elora (6.7 pH, CEC 21 meq/100g) and KUBC (pH 5.6, CEC 28 meq/100g) with higher pH and CEC only lost 13.6% and 12.2% total metals respectively. Metal losses were highest for Ni, Zn and Co (46.0%, 63.7% and 48.4% metal loss respectively) whereas Pb and Cu (5.6% and 20.0% metal loss respectively) were mostly retained, affecting mixture ratios. Comparatively, oxide and annealed metal dosing which do not require leaching had higher total metal concentrations, closer to nominal doses and maintained better mixture ratios (percent of nominal concentrations for the oxide metal dosing were Pb = 109.9%, Cu = 84.6%, Ni = 101.9%, Zn = 82.3% and Co = 97.8% and for the annealed metal dosing were Pb = 81.7%, Cu = 80.3%, Ni = 100.5%, Zn = 89.2% and Co = 101.3%). Relative to their total metal concentrations, nitrate metal dosing (lowest metal concentrations) was the most toxic followed by metal oxides dosing while the annealed dosing method was generally non-toxic. Due to the lack of toxicity of the annealed metals and their higher dosing effort, metal oxides, are the most appropriate of the tested dosing methods, for fixed-ratio metal mixtures studies with soil invertebrates.
Sigma-Aldrich
Nickel(II) oxide, green, −325 mesh, 99%
Sigma-Aldrich
Lead(II) oxide, ACS reagent, ≥99.0%
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Copper(II) nitrate hemi(pentahydrate), ACS reagent, 98%
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, purum p.a., crystallized, ≥97.0% (KT)
Sigma-Aldrich
Zinc nitrate hexahydrate, reagent grade, 98%
Sigma-Aldrich
Cobalt(II,III) oxide, powder, <10 μm
Sigma-Aldrich
Copper(II) oxide, powder, <10 μm, 98%
Sigma-Aldrich
Cobalt(II) nitrate hexahydrate, reagent grade, 98%
Sigma-Aldrich
Lead(II) nitrate, ACS reagent, ≥99.0%