Merck
  • Rational design of thiolated polyenes as trifunctional Raman reporter molecules in surface-enhanced Raman scattering nanotags for cytokine detection in a lateral flow assay.
Rational design of thiolated polyenes as trifunctional Raman reporter molecules in surface-enhanced Raman scattering nanotags for cytokine detection in a lateral flow assay.
Journal of biophotonics (2020-01-21)
Thomas Keller, Svetlana Brem, Vi Tran, Oliver Sritharan, Daniel Schäfer, Sebastian Schlücker
ABSTRACT
The characteristic vibrational spectroscopic fingerprint of Raman reporter molecules adsorbed on noble metal nanoparticles is employed for the identification of target proteins by the corresponding surface-enhanced Raman scattering (SERS) nanotag-labeled antibodies. Here, we present the modular synthesis of thiolated polyenes with two to five C═C double bonds introduced via stepwise Wittig reactions. The experimental characterization of their electronic and vibrational properties is complemented by density functional theory calculations. Highly SERS-active nanotags are generated by using the thiolated polyenes as Raman reporter molecules in Au/Au core/satellite supraparticles with multiple hot spots. The cytokines IL-1β and IFN-γ are detected in a duplex SERS-based lateral flow assay on a nitrocellulose test strip by Raman microscopy. The thiolated polyenes are suitable for use in immuno-SERS applications such as point-of-care testing as well as cellular and tissue imaging.
Sigma-Aldrich
Sodium bromide, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
4-Bromobenzyl alcohol, 99%
Sigma-Aldrich
Cyclohexane, Laboratory Reagent, ≥99.8%
Sigma-Aldrich
(11-Mercaptoundecyl)-N,N,N-trimethylammonium bromide
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
4-(Methylthio)benzaldehyde, 95%
Sigma-Aldrich
2-Methyl-2-propanethiol, 99%
Sigma-Aldrich
Ethyl bromoacetate, reagent grade, 98%
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%