Merck
  • A case-control study of perfluoroalkyl substances and the risk of breast cancer in Taiwanese women.
A case-control study of perfluoroalkyl substances and the risk of breast cancer in Taiwanese women.
Environment international (2020-06-25)
Meng-Shan Tsai, Shu-Han Chang, Wen-Hung Kuo, Ching-Hua Kuo, Szu-Yi Li, Ming-Yang Wang, Dwan-Ying Chang, Yen-Shen Lu, Chiun-Sheng Huang, Ann-Lii Cheng, Ching-Hung Lin, Pau-Chung Chen
ABSTRACT
Breast cancer (BC) is a common cancer in women worldwide; however, the incidence of BC is increasing in younger women, possibly associated with the environment. Perfluoroalkyl substances (PFAS) are one of endocrine disruptors that accumulate in environment and impact human health. This study aimed to investigate whether the PFAS and BC are associated. We enrolled 120 BCE patients and 119 controls at National Taiwan University Hospital (NTUH) and also collected bio-specimen and questionnaire from 2013 to 2015. All subjects' plasma PFAS levels were analyzed by ultra-performance liquid chromatography tandem mass spectrometry method with electrospray ionization (UHPLC-ESI-MS/MS). A logistic regression model was used to estimate the association between PFAS and BC. In the ≤50 years age group, the adjusted odds ratio (OR) was 2.34 (95% CI = 1.02, 5.38) for perfluorooctane sulfonate (PFOS) exposure per natural log unit increase. After stratifying the estrogen receptor (ER) status and age group, we obtained a positive association for PFHxS and PFOS concentrations with respect to the risk of ER positive tumors for ≤50 years age group. In conclusion, we found that PFAS were associated with the BC risk of ER positive tumors in young Taiwanese women. Further studies are needed to follow and explore whether these associations are causal.
Sigma-Aldrich
Perfluoroheptanoic acid, ≥97.0%
Supelco
Pentadecafluorooctanoic acid solution, 100 μg/mL in methanol, analytical standard
Sigma-Aldrich
Perfluorodecanoic acid, 97%