Merck
  • Altered immune function during long-term host-tumor interactions can be modulated to retard autochthonous neoplastic growth.
Altered immune function during long-term host-tumor interactions can be modulated to retard autochthonous neoplastic growth.
Journal of immunology (Baltimore, Md. : 1950) (2007-08-22)
Trina J Stewart, Scott I Abrams
ABSTRACT
Ag-specific and generalized forms of immunosuppression have been documented in animal tumor models. However, much of our knowledge on tumor-induced immunosuppression was acquired using tumor implant models, which do not reiterate the protracted nature of host-tumor interactions. Therefore, a transgenic mouse model of autochthonous mammary tumor development and progression was chosen to investigate the long-term consequences of neoplastic growth on the immune system. In vitro proliferation of unfractionated splenocytes from tumor-bearing mice, as assessed by [(3)H]thymidine uptake, was inhibited by the presence of suppressor cells within these splenocyte preparations, because purifying the T cells restored their biological activity. However, the level of inhibition did not correlate with either tumor load or the percentage of myeloid-derived CD11b+Gr1+ cells. To evaluate tumor-specific immune dysfunction, transgenic mice were challenged with autologous tumor cells. Mice with extensive, but not minimal autochthonous tumor burdens demonstrated a significantly enhanced rate of autologous tumor growth compared with age-matched controls. In contrast, an allogeneic tumor challenge was efficiently rejected from both groups of transgenic mice. It was also noted that allogeneic tumor challenge of mice with minimal disease significantly inhibited autochthonous primary tumor growth. We therefore demonstrated that 1) a generalized form of immunosuppression occurred, but not as a result of permanent alterations to T cell function, because purified T cell subsets retained normal biological activity following polyclonal or allostimulation; and 2) tumor-specific immunosuppression emerged as a consequence of tumor progression, but could be modulated to enhance antitumor responses against autochthonous primary neoplastic growth.
Sigma-Aldrich
AT-3 Mouse Mammary Carcinoma Cell Line, AT-3 mouse mammary tumor cell line may be used to develop mouse models for mammary cancer studies.