Merck
  • RIM and RIM-BP Form Presynaptic Active-Zone-like Condensates via Phase Separation.
RIM and RIM-BP Form Presynaptic Active-Zone-like Condensates via Phase Separation.
Molecular cell (2019-01-22)
Xiandeng Wu, Qixu Cai, Zeyu Shen, Xudong Chen, Menglong Zeng, Shengwang Du, Mingjie Zhang
ABSTRACT
Both the timing and kinetics of neurotransmitter release depend on the positioning of clustered Ca2+ channels in active zones to docked synaptic vesicles on presynaptic plasma membranes. However, how active zones form is not known. Here, we show that RIM and RIM-BP, via specific multivalent bindings, form dynamic and condensed assemblies through liquid-liquid phase separation. Voltage-gated Ca2+ channels (VGCCs), via C-terminal-tail-mediated direct binding to both RIM and RIM-BP, can be enriched to the RIM and RIM-BP condensates. We further show that RIM and RIM-BP, together with VGCCs, form dense clusters on the supported lipid membrane bilayers via phase separation. Therefore, RIMs and RIM-BPs are plausible organizers of active zones, and the formation of RIM and RIM-BP condensates may cluster VGCCs into nano- or microdomains and position the clustered Ca2+ channels with Ca2+ sensors on docked vesicles for efficient and precise synaptic transmissions.
Product Description
Avanti
18:0 Cyanine 5 PE, Avanti Research - A Croda Brand
Avanti
18:1 Cyanine 5 PE, Avanti Research - A Croda Brand
Avanti
18:2 TopFluor cholesterol, Avanti Research - A Croda Brand 810290P, powder