Merck
  • Accurate mass and retention time library of serum lipids for type 1 diabetes research.
Accurate mass and retention time library of serum lipids for type 1 diabetes research.
Analytical and bioanalytical chemistry (2019-07-08)
Ngoc Vu, Monica Narvaez-Rivas, Guan-Yuan Chen, Marian J Rewers, Qibin Zhang
ABSTRACT
Dysregulated lipid species are linked to various disease pathologies and implicated as potential biomarkers for type 1 diabetes (T1D). However, it is challenging to comprehensively profile the blood specimen lipidome with full structural details of every lipid molecule. The commonly used reversed-phase liquid chromatography-tandem mass spectrometry (RPLC-MS/MS)-based lipidomics approach is powerful for the separation of individual lipid species, but lipids belonging to different classes may still co-elute and result in ion suppression and misidentification of lipids. Using offline mixed-mode and RPLC-based two-dimensional separations coupled with MS/MS, a comprehensive lipidomic profiling was performed on human sera pooled from healthy and T1D subjects. The elution order of lipid molecular species on RPLC showed good correlations to the total number of carbons in fatty acyl chains and total number of double bonds. This observation together with fatty acyl methyl ester analysis was used to enhance the confidence of identified lipid species. The final T1D serum lipid library database contains 753 lipid molecular species with accurate mass and RPLC retention time uniquely annotated for each of the species. This comprehensive human serum lipid library can serve as a database for high-throughput RPLC-MS-based lipidomic analysis of blood samples related to T1D and other childhood diseases. Graphical abstract.
Product Description
Avanti
d5-TG ISTD Mix I, Avanti Research - A Croda Brand LM6000
Avanti
Sphingomyelin (d18:1/12:0), Avanti Research - A Croda Brand LM2312, ethanol solution